Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ALOX15B Antibodies

          AbboMax

          15-Lox-2 Polyclonal Antibody

          View all (9) ALOX15B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite 15-Lox-2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          15-Lox-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          15-Lox-2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          15-Lox-2 Antibody (600-590) in WB

          Western Blot: The whole lysate derived from HT29 (20 µg/lane) immunoblotted by Rabbit anti – 15-LOX2 (Cat#600-590) at 1:500. Observed a major immunoreactive band at molecular weight ~74 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          15-Lox-2 Antibody in Western Blot (WB)
          15-Lox-2 Polyclonal Antibody

          Product Details

          600-590

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide LP YTY LDP PLI ENS VSI corresponding to the internal portion of human LOX2 protein sequence
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with proprietary stabilizer

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Positive control: HT-29 cells

          Cellular location: Cytoplasmic.

          Target Information

          ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15-LOX2sv-a; 15-LOX2sv-b; 15-LOX2sv-c; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; arachidonate 15-lipoxygenase 2; Arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; Arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; Linoleate 13-lipoxygenase 15-LOb; Linoleate 9S-lipoxygenase ALOX8; Polyunsaturated fatty acid lipoxygenase ALOX15B; Polyunsaturated fatty acid lipoxygenase ALOX8; unnamed protein product

          View more View less

          Gene Aliases: 15-LOX-2; 15-LOX-B; 8-LOX; 8S-LOX; ALOX15B; Alox8

          View more View less

          UniProt ID: (Human) O15296, (Rat) Q8K4F2, (Mouse) O35936

          View more View less

          Entrez Gene ID: (Human) 247, (Rat) 266604, (Mouse) 11688

          View more View less

          Function(s)
          iron ion binding calcium ion binding protein binding lipid binding linoleate 13S-lipoxygenase activity oxidoreductase activity oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen arachidonate 8(S)-lipoxygenase activity metal ion binding arachidonate 15-lipoxygenase activity dioxygenase activity linoleate 9S-lipoxygenase activity 2,3-dihydroxy DDT 1,2-dioxygenase activity phenanthrene dioxygenase activity 2,2',3-trihydroxybiphenyl dioxygenase activity 1,2-dihydroxyfluorene 1,1-alpha-dioxygenase activity 5,6-dihydroxy-3-methyl-2-oxo-1,2-dihydroquinoline dioxygenase activity 1,1-dichloro-2-(dihydroxy-4-chlorophenyl)-(4-chlorophenyl)ethene 1,2-dioxygenase activity protocatechuate 3,4-dioxygenase type II activity 2'-aminobiphenyl-2,3-diol 1,2-dioxygenase activity 3,4-dihydroxyfluorene 4,4-alpha-dioxygenase activity 2,3-dihydroxy-ethylbenzene 1,2-dioxygenase activity carbazole 1,9a-dioxygenase activity dihydroxydibenzothiophene dioxygenase activity 1,2-dihydroxynaphthalene-6-sulfonate 1,8a-dioxygenase activity styrene dioxygenase activity 3,4-dihydroxyphenanthrene dioxygenase activity hydroquinone 1,2-dioxygenase activity p-cumate 2,3-dioxygenase activity 2,3-dihydroxy-p-cumate dioxygenase activity 3,5-dichlorocatechol 1,2-dioxygenase activity 2-aminophenol 1,6-dioxygenase activity 2,6-dichloro-p-hydroquinone 1,2-dioxygenase activity chlorocatechol 1,2-dioxygenase activity catechol dioxygenase activity dihydroxyfluorene dioxygenase activity 5-aminosalicylate dioxygenase activity 3-hydroxy-2-naphthoate 2,3-dioxygenase activity benzo(a)pyrene 11,12-dioxygenase activity benzo(a)pyrene 4,5-dioxygenase activity 4,5-dihydroxybenzo(a)pyrene dioxygenase activity benzo(a)pyrene 9,10-dioxygenase activity 9,10-dihydroxybenzo(a)pyrene dioxygenase activity benzo(a)pyrene 7,8-dioxygenase activity 7,8-dihydroxy benzo(a)pyrene dioxygenase activity 1,2-dihydroxy-5,6,7,8-tetrahydronaphthalene extradiol dioxygenase activity 2-mercaptobenzothiazole dioxygenase activity pyridine-3,4-diol dioxygenase activity pyrene dioxygenase activity 4,5-dihydroxypyrene dioxygenase activity phenanthrene-4-carboxylate dioxygenase activity tetrachlorobenzene dioxygenase activity 4,6-dichloro-3-methylcatechol 1,2-dioxygenase activity 2,3-dihydroxydiphenyl ether dioxygenase activity diphenyl ether 1,2-dioxygenase activity 4-hydroxycatechol 1,2-dioxygenase activity arachidonate 8(R)-lipoxygenase activity oxygenase
          Process(es)
          lipid metabolic process phospholipid metabolic process apoptotic process negative regulation of cell proliferation positive regulation of macrophage derived foam cell differentiation arachidonic acid metabolic process lipoxygenase pathway negative regulation of cell migration prostate gland development regulation of epithelial cell differentiation positive regulation of chemokine production lipid oxidation positive regulation of peroxisome proliferator activated receptor signaling pathway linoleic acid metabolic process positive regulation of keratinocyte differentiation negative regulation of cell cycle negative regulation of growth hepoxilin biosynthetic process endocannabinoid signaling pathway cannabinoid biosynthetic process lipoxin A4 biosynthetic process oxidation-reduction process positive regulation of chemokine secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0