    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD301b Antibodies
          Invitrogen
          CD301b (MGL2) Monoclonal Antibody (11A10-B7), APC, eBioscience™
          View all (3) CD301b antibodies
          Questions & Answers
          Cite CD301b (MGL2) Monoclonal Antibody (11A10-B7), APC, eBioscience™
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CD301b (MGL2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD301b (MGL2) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD301b (MGL2) Antibody (17-3011-82) in Flow
          C57BL/6 mouse skin draining lymph node cells were stained with CD3 Monoclonal Antibody, FITC (Product # 11-0032-82), CD19 Monoclonal Antibody, FITC (Product # 11-0193-82), MHC Class II (I-A/I-E) Monoclonal Antibody, eFluor 450 (Product # 48-5321-82), CD11c Monoclonal Antibody, PE (Product # 12-0114-82) and 0.125 µg of Rat IgG2a kappa Isotype Control, APC (Product # 17-4321-81) (left) or 0.125 µg of CD301b (MGL2) Monoclonal Antibody, APC (right). Viable cells, as determined by 7-AAD (Product # 00-6993-50), that were CD19-, CD3-, MHC II... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD301b (MGL2) Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          17-3011-82
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg/test
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rat / IgG2a, lambda
          Recommended Isotype Control
          Rat IgG2a kappa Isotype Control (eBR2a), APC, eBioscience™
          Class
          Monoclonal
          Type
          Antibody
          Clone
          11A10-B7
          Conjugate
          APC APC APC
          • Unconjugated
          • eFluor 660
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          liquid
          Concentration
          0.2 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Description
          The monoclonal antibody 11A10-B7 recognizes mouse CD301b. CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.
          Applications Tested
          This 11A10-B7 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 0.25 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Staining Protocol: All protocols work well for this monoclonal antibody. Use of Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol: intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to "Clone Performance Following Fixation/Permeabilization" located in the BestProtocols Section under the Resources tab online). All Protocols can be found in the Flow Cytometry Protocols: "Staining Intracellular Antigens for Flow Cytometry Protocol" located in the BestProtocols®Section under the Resources tab online.
          Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
          Target Information
          CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD301; macrophage galactose-type C-type lectin 2; mMGL2i; lectin; unnamed protein product
          View more View less
          Gene Aliases: CD301b
          View more View less
          Entrez Gene ID: (Mouse) 216864
          View more View less
          Function(s)
          mannose binding carbohydrate binding pattern recognition receptor activity fucose binding
          Process(es)
          immune response
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch