Product References
DOT1L Is a Novel Cancer Stem Cell Target for Triple-Negative Breast Cancer.
Clinical cancer research : an official journal of the American Association for Cancer Research
Kurani H,Razavipour SF,Harikumar KB,Dunworth M,Ewald AJ,Nasir A,Pearson G,Van Booven D,Zhou Z,Azzam D,Wahlestedt C,Slingerland J
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Mon May 02 00:00:00 EDT 2022
Astrocytic interleukin-3 programs microglia and limits Alzheimer's disease.
Nature
McAlpine CS,Park J,Griciuc A,Kim E,Choi SH,Iwamoto Y,Kiss MG,Christie KA,Vinegoni C,Poller WC,Mindur JE,Chan CT,He S,Janssen H,Wong LP,Downey J,Singh S,Anzai A,Kahles F,Jorfi M,Feruglio PF,Sadreyev RI,Weissleder R,Kleinstiver BP,Nahrendorf M,Tanzi RE,Swirski FK
17-5071-42 was used in Flow cytometry/Cell sorting to identify IL-3 as a key mediator of astrocyte-microglia cross-talk and a node for therapeutic intervention in Alzheimer's disease.
Thu Jul 01 00:00:00 EDT 2021
Gain-of-function hot spot mutant p53R248Q regulation of integrin/FAK/ERK signaling in esophageal squamous cell carcinoma.
Translational oncology
Yu VZ,So SS,Lung ML
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Gain-of-function hot spot mutant p53R248Q regulation of integrin/FAK/ERK signaling in esophageal squamous cell carcinoma.
Translational oncology
Yu VZ,So SS,Lung ML
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
BCOR Internal Tandem Duplication Expression in Neural Stem Cells Promotes Growth, Invasion, and Expression of PRC2 Targets.
International journal of molecular sciences
Nakata S,Yuan M,Rubens JA,Kahlert UD,Maciaczyk J,Raabe EH,Eberhart CG
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Sat Apr 10 00:00:00 EDT 2021
IgA Plasma Cells Are Long-Lived Residents of Gut and Bone Marrow That Express Isotype- and Tissue-Specific Gene Expression Patterns.
Frontiers in immunology
Wilmore JR,Gaudette BT,Gómez Atria D,Rosenthal RL,Reiser SK,Meng W,Rosenfeld AM,Luning Prak ET,Allman D
17-5071-42 was used in Flow Cytometry to suggest that systemic and mucosal IgA+ plasma cells are from a common source, but they do not migrate between tissues.
Wed Feb 16 00:00:00 EST 2022
PD-L1 recruits phospholipase C and enhances tumorigenicity of lung tumors harboring mutant forms of EGFR.
Cell reports
Ghosh S,Nataraj NB,Noronha A,Patkar S,Sekar A,Mukherjee S,Winograd-Katz S,Kramarski L,Verma A,Lindzen M,Garcia DD,Green J,Eisenberg G,Gil-Henn H,Basu A,Lender Y,Weiss S,Oren M,Lotem M,Geiger B,Ruppin E,Yarden Y
17-5071-42 was used in Immunocytochemistry to portray PD-L1 as a molecular amplifier of EGFR signaling and improve the understanding of the resistance of EGFR+ tumors to immunotherapy.
Tue May 25 00:00:00 EDT 2021
Detecting chromatin interactions between and along sister chromatids with SisterC.
Nature methods
Oomen ME,Hedger AK,Watts JK,Dekker J
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Myofiber necroptosis promotes muscle stem cell proliferation via releasing Tenascin-C during regeneration.
Cell research
Zhou S,Zhang W,Cai G,Ding Y,Wei C,Li S,Yang Y,Qin J,Liu D,Zhang H,Shao X,Wang J,Wang H,Yang W,Wang H,Chen S,Hu P,Sun L
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Tue Dec 01 00:00:00 EST 2020
Insulin-Like Growth Factors Are Key Regulators of T Helper 17 Regulatory T Cell Balance in Autoimmunity.
Immunity
DiToro D,Harbour SN,Bando JK,Benavides G,Witte S,Laufer VA,Moseley C,Singer JR,Frey B,Turner H,Bruning J,Darley-Usmar V,Gao M,Conover C,Hatton RD,Frank S,Colonna M,Weaver CT
17-5071 was used in Flow cytometry/Cell sorting to show that, among effector T cell subsets, Th17 and Treg cells selectively expressed multiple components of the IGF system.
Tue Apr 14 00:00:00 EDT 2020
Prostate Epithelial RON Signaling Promotes M2 Macrophage Activation to Drive Prostate Tumor Growth and Progression.
Molecular cancer research : MCR
Sullivan C,Brown NE,Vasiliauskas J,Pathrose P,Starnes SL,Waltz SE
17-5071 was used in Flow cytometry/Cell sorting to provide additional evidence that epithelial RON expression coordinates interactions between prostate tumor cells and macrophages to promote macrophage-mediated tumor cell growth.
Sat Aug 01 00:00:00 EDT 2020
Autotaxin inhibition reduces cardiac inflammation and mitigates adverse cardiac remodeling after myocardial infarction.
Journal of molecular and cellular cardiology
Tripathi H,Al-Darraji A,Abo-Aly M,Peng H,Shokri E,Chelvarajan L,R Donahue R,Levitan BM,Gao E,Hernandez G,Morris AJ,Smyth SS,Abdel-Latif A
17-5071-42 was used in Flow Cytometry to study the autotaxin activity and lysophosphatidic acid levels in relation to cardiac and systemic inflammation in acute myocardial infarction patients and C57BL/6 (WT) mice.
Tue Dec 01 00:00:00 EST 2020
ASH2L drives proliferation and sensitivity to bleomycin and other genotoxins in Hodgkin's lymphoma and testicular cancer cells.
Cell death & disease
Constantin D,Widmann C
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Mon Nov 30 00:00:00 EST 2020
Differential expansion of T central memory precursor and effector subsets is regulated by division speed.
Nature communications
Kretschmer L,Flossdorf M,Mir J,Cho YL,Plambeck M,Treise I,Toska A,Heinzel S,Schiemann M,Busch DH,Buchholz VR
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Wed Jan 08 00:00:00 EST 2020
Sleep modulates haematopoiesis and protects against atherosclerosis.
Nature
McAlpine CS,Kiss MG,Rattik S,He S,Vassalli A,Valet C,Anzai A,Chan CT,Mindur JE,Kahles F,Poller WC,Frodermann V,Fenn AM,Gregory AF,Halle L,Iwamoto Y,Hoyer FF,Binder CJ,Libby P,Tafti M,Scammell TE,Nahrendorf M,Swirski FK
17-5071-42 was used in Flow Cytometry to investigate the cellular and molecular mechanisms by which sleep maintains cardiovascular health.
Fri Feb 01 00:00:00 EST 2019
The H3K4 methyltransferase Setd1b is essential for hematopoietic stem and progenitor cell homeostasis in mice.
eLife
Schmidt K,Zhang Q,Tasdogan A,Petzold A,Dahl A,Arneth BM,Slany R,Fehling HJ,Kranz A,Stewart AF,Anastassiadis K
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Tue Jun 19 00:00:00 EDT 2018
FOXP1 controls mesenchymal stem cell commitment and senescence during skeletal aging.
The Journal of clinical investigation
Li H,Liu P,Xu S,Li Y,Dekker JD,Li B,Fan Y,Zhang Z,Hong Y,Yang G,Tang T,Ren Y,Tucker HO,Yao Z,Guo X
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Mon Apr 03 00:00:00 EDT 2017
Loss of Forkhead box M1 promotes erythropoiesis through increased proliferation of erythroid progenitors.
Haematologica
Youn M,Wang N,LaVasseur C,Bibikova E,Kam S,Glader B,Sakamoto KM,Narla A
17-5071 was used in Flow cytometry/Cell sorting to find that inhibition of FOXM1, forkhead domain inhibitor-6, increases red blood cell numbers through additional proliferation of erythroid precursors.
Mon May 01 00:00:00 EDT 2017
4PD Functionalized Dendrimers: A Flexible Tool for In Vivo Gene Silencing of Tumor-Educated Myeloid Cells.
Journal of immunology (Baltimore, Md. : 1950)
Zilio S,Vella JL,De la Fuente AC,Daftarian PM,Weed DT,Kaifer A,Marigo I,Leone K,Bronte V,Serafini P
17-5071 was used in Flow cytometry/Cell sorting to study the use of 4PD nanoparticles for the in vivo gene silencing of tumour-educated myeloid cells.
Mon May 15 00:00:00 EDT 2017
PKN2 is essential for mouse embryonic development and proliferation of mouse fibroblasts.
Genes to cells : devoted to molecular & cellular mechanisms
Danno S,Kubouchi K,Mehruba M,Abe M,Natsume R,Sakimura K,Eguchi S,Oka M,Hirashima M,Yasuda H,Mukai H
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2017
Downregulation of ZBTB24 hampers the G0/1- to S-phase cell-cycle transition via upregulating the expression of IRF-4 in human B cells.
Genes and immunity
Liang J,Yan R,Chen G,Feng J,Wu WW,Ren W,Zhu C,Zhao Y,Gao XM,Wang J
17-5071-42 was used in Flow Cytometry to show that downregulation of ZBTB24 hampers the G0/1- to S-phase cell-cycle transition via upregulating the expression of IRF-4 in human B cells.
Fri Jul 01 00:00:00 EDT 2016
Breast-cancer-secreted miR-122 reprograms glucose metabolism in premetastatic niche to promote metastasis.
Nature cell biology
Fong MY,Zhou W,Liu L,Alontaga AY,Chandra M,Ashby J,Chow A,O'Connor ST,Li S,Chin AR,Somlo G,Palomares M,Li Z,Tremblay JR,Tsuyada A,Sun G,Reid MA,Wu X,Swiderski P,Ren X,Shi Y,Kong M,Zhong W,Chen Y,Wang SE
Sun Feb 01 00:00:00 EST 2015
Functional genomic screening reveals asparagine dependence as a metabolic vulnerability in sarcoma.
eLife
Hettmer S,Schinzel AC,Tchessalova D,Schneider M,Parker CL,Bronson RT,Richards NG,Hahn WC,Wagers AJ
17-5071 was used in Flow cytometry/Cell sorting to investigate gene targets by selective targeting of the molecular networks that support sarcoma cell proliferation.
Sat Oct 24 00:00:00 EDT 2015
CENP-32 is required to maintain centrosomal dominance in bipolar spindle assembly.
Molecular biology of the cell
Ohta S,Wood L,Toramoto I,Yagyu K,Fukagawa T,Earnshaw WC
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
Programmed death 1 regulates memory phenotype CD4 T cell accumulation, inhibits expansion of the effector memory phenotype subset and modulates production of effector cytokines.
PloS one
Charlton JJ,Tsoukatou D,Mamalaki C,Chatzidakis I
17-5071 was used in Flow cytometry/Cell sorting to demonstrate that PD-1 has an important role in determining the composition and functional aspects of the memory phenotype CD4 T cell pool.
Wed Apr 13 00:00:00 EDT 2016
Atm deletion with dual recombinase technology preferentially radiosensitizes tumor endothelium.
The Journal of clinical investigation
Moding EJ,Lee CL,Castle KD,Oh P,Mao L,Zha S,Min HD,Ma Y,Das S,Kirsch DG
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Interleukin-7 expression and its effect on natural killer cells in patients with multiple sclerosis.
Journal of neuroimmunology
Su N,Shi SX,Zhu X,Borazanci A,Shi FD,Gan Y
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Sat Nov 15 00:00:00 EST 2014
MiR-630 inhibits proliferation by targeting CDC7 kinase, but maintains the apoptotic balance by targeting multiple modulators in human lung cancer A549 cells.
Cell death & disease
Cao JX,Lu Y,Qi JJ,An GS,Mao ZB,Jia HT,Li SY,Ni JH
Published figure using BrdU monoclonal antibody (Product # 17-5071-42) in Flow Cytometry
Thu Sep 25 00:00:00 EDT 2014
Soluble factors secreted by T cells promote β-cell proliferation.
Diabetes
Dirice E,Kahraman S,Jiang W,El Ouaamari A,De Jesus DF,Teo AK,Hu J,Kawamori D,Gaglia JL,Mathis D,Kulkarni RN
17-5071 was used in Immunohistochemistry to study the role of soluble factors secreted from T cells to enhance pancreatic β-cell proliferation.
Wed Jan 01 00:00:00 EST 2014
Ponatinib suppresses the development of myeloid and lymphoid malignancies associated with FGFR1 abnormalities.
Leukemia
Ren M,Qin H,Ren R,Cowell JK
17-5071 was used in Flow cytometry/Cell sorting to investigate the use of ponatinib for treatment of myeloid and lymphoid malignancies.
Tue Jan 01 00:00:00 EST 2013
Selective reduction of post-selection CD8 thymocyte proliferation in IL-15Rα deficient mice.
PloS one
Chow KP,Qiu JT,Lee JM,Hsu SL,Yang SC,Wu NN,Huang W,Wu TS
17-5071 was used in Flow cytometry/Cell sorting to investigate the unique IL-15-dependent cell expansion at the mature CD8SP stage of thymocyte development.
Mon Aug 20 00:00:00 EDT 2012