Product References
MiR-451a and let-7i-5p loaded extracellular vesicles attenuate heme-induced inflammation in hiPSC-derived endothelial cells.
Frontiers in immunology
Thomas JJ,Harp KO,Bashi A,Hood JL,Botchway F,Wilson MD,Thompson WE,Stiles JK,Driss A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Wed Jan 11 00:00:00 EST 2023
Angiogenic stem cell delivery platform to augment post-infarction neovasculature and reverse ventricular remodeling.
Scientific reports
Shin HS,Thakore A,Tada Y,Pedroza AJ,Ikeda G,Chen IY,Chan D,Jaatinen KJ,Yajima S,Pfrender EM,Kawamura M,Yang PC,Wu JC,Appel EA,Fischbein MP,Woo Y,Shudo Y
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Thu Oct 20 00:00:00 EDT 2022
Expression and role of interleukin-1β and associated biomarkers in deep vein thrombosis.
Experimental and therapeutic medicine
Pai RZ,Fang Q,Tian G,Zhu B,Ge X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Wed Dec 01 00:00:00 EST 2021
Injectable hydrogel with MSNs/microRNA-21-5p delivery enables both immunomodification and enhanced angiogenesis for myocardial infarction therapy in pigs.
Science advances
Li Y,Chen X,Jin R,Chen L,Dang M,Cao H,Dong Y,Cai B,Bai G,Gooding JJ,Liu S,Zou D,Zhang Z,Yang C
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Down-Regulated Exosomal MicroRNA-221 - 3p Derived From Senescent Mesenchymal Stem Cells Impairs Heart Repair.
Frontiers in cell and developmental biology
Sun L,Zhu W,Zhao P,Zhang J,Lu Y,Zhu Y,Zhao W,Liu Y,Chen Q,Zhang F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Mon Sep 28 00:00:00 EDT 2020
Aging affects responsiveness of peripheral blood mononuclear cells to immunosuppression of periodontal ligament stem cells.
The Journal of international medical research
Li X,Zhang B,Wang H,Zhao X,Zhang Z,Ding G,Wei F
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Wed Jul 01 00:00:00 EDT 2020
PF-127 hydrogel plus sodium ascorbyl phosphate improves Wharton's jelly mesenchymal stem cell-mediated skin wound healing in mice.
Stem cell research & therapy
Deng Q,Huang S,Wen J,Jiao Y,Su X,Shi G,Huang J
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Fri Apr 03 00:00:00 EDT 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Nov 17 00:00:00 EST 2020
Both Human Hematoma Punctured from Pelvic Fractures and Serum Increase Muscle Resident Stem Cells Response to BMP9: A Multivariate Statistical Approach.
Journal of clinical medicine
Alinejad Y,Lauzon MA,Grenier G,Balg F,Faucheux N
25-0319-42 was used in Flow Cytometry to study the influence of Hema, human healthy serum, and fetal bovine serum on bone morphogenetic protein 9 (BMP9)-induced osteoblast commitment of muscle resident mesenchymal stromal cells by measuring alkaline phosphatase activity.
Sun Apr 19 00:00:00 EDT 2020
Both Human Hematoma Punctured from Pelvic Fractures and Serum Increase Muscle Resident Stem Cells Response to BMP9: A Multivariate Statistical Approach.
Journal of clinical medicine
Alinejad Y,Lauzon MA,Grenier G,Balg F,Faucheux N
25-0319-42 was used in Flow Cytometry to study the influence of Hema, human healthy serum, and fetal bovine serum on bone morphogenetic protein 9 (BMP9)-induced osteoblast commitment of muscle resident mesenchymal stromal cells by measuring alkaline phosphatase activity.
Sun Apr 19 00:00:00 EDT 2020
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Functionally specialized human CD4+ T-cell subsets express physicochemically distinct TCRs.
eLife
Kasatskaya SA,Ladell K,Egorov ES,Miners KL,Davydov AN,Metsger M,Staroverov DB,Matveyshina EK,Shagina IA,Mamedov IZ,Izraelson M,Shelyakin PV,Britanova OV,Price DA,Chudakov DM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Dec 08 00:00:00 EST 2020
Vascular Tumor Recapitulated in Endothelial Cells from hiPSCs Engineered to Express the SERPINE1-FOSB Translocation.
Cell reports. Medicine
van IJzendoorn DGP,Salvatori DCF,Cao X,van den Hil F,Briaire-de Bruijn IH,de Jong D,Mei H,Mummery CL,Szuhai K,Bovée JVMG,Orlova VV
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Dec 22 00:00:00 EST 2020
Spatial and Single-Cell Transcriptional Profiling Identifies Functionally Distinct Human Dermal Fibroblast Subpopulations.
The Journal of investigative dermatology
Philippeos C,Telerman SB,Oulès B,Pisco AO,Shaw TJ,Elgueta R,Lombardi G,Driskell RR,Soldin M,Lynch MD,Watt FM
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Expansion of functional personalized cells with specific transgene combinations.
Nature communications
Lipps C,Klein F,Wahlicht T,Seiffert V,Butueva M,Zauers J,Truschel T,Luckner M,Köster M,MacLeod R,Pezoldt J,Hühn J,Yuan Q,Müller PP,Kempf H,Zweigerdt R,Dittrich-Breiholz O,Pufe T,Beckmann R,Drescher W,Riancho J,Sañudo C,Korff T,Opalka B,Rebmann V,Göthert JR,Alves PM,Ott M,Schucht R,Hauser H,Wirth D,May T
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Thu Mar 08 00:00:00 EST 2018
Prostaglandin E2 Is Required for BMP4-Induced Mesoderm Differentiation of Human Embryonic Stem Cells.
Stem cell reports
Zhang B,He L,Liu Y,Zhang J,Zeng Q,Wang S,Fan Z,Fang F,Chen L,Lv Y,Xi J,Yue W,Li Y,Pei X
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Mar 13 00:00:00 EDT 2018
Functionally distinct disease-associated fibroblast subsets in rheumatoid arthritis.
Nature communications
Mizoguchi F,Slowikowski K,Wei K,Marshall JL,Rao DA,Chang SK,Nguyen HN,Noss EH,Turner JD,Earp BE,Blazar PE,Wright J,Simmons BP,Donlin LT,Kalliolias GD,Goodman SM,Bykerk VP,Ivashkiv LB,Lederer JA,Hacohen N,Nigrovic PA,Filer A,Buckley CD,Raychaudhuri S,Brenner MB
25-0319 was used in Flow cytometry/Cell sorting to identify seven fibroblast subsets with distinct surface protein phenotypes, and characterise them into three subsets by integrating transcriptomic data.
Fri Feb 23 00:00:00 EST 2018
Stromal cell cadherin-11 regulates adipose tissue inflammation and diabetes.
The Journal of clinical investigation
Chang SK,Kohlgruber AC,Mizoguchi F,Michelet X,Wolf BJ,Wei K,Lee PY,Lynch L,Duquette D,Ceperuelo-Mallafré V,Banks AS,Brenner MB
25-0319 was used in Flow cytometry/Cell sorting to suggest that stromal fibroblasts expressing cadherin-11 regulate adipose tissue inflammation.
Fri Sep 01 00:00:00 EDT 2017
MMP-2 and MMP-14 Silencing Inhibits VEGFR2 Cleavage and Induces the Differentiation of Porcine Adipose-Derived Mesenchymal Stem Cells to Endothelial Cells.
Stem cells translational medicine
Almalki SG,Llamas Valle Y,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Mon May 01 00:00:00 EDT 2017
ERK signaling is required for VEGF-A/VEGFR2-induced differentiation of porcine adipose-derived mesenchymal stem cells into endothelial cells.
Stem cell research & therapy
Almalki SG,Agrawal DK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Fri May 12 00:00:00 EDT 2017
TGF-β reduces DNA ds-break repair mechanisms to heighten genetic diversity and adaptability of CD44+/CD24- cancer cells.
eLife
Pal D,Pertot A,Shirole NH,Yao Z,Anaparthy N,Garvin T,Cox H,Chang K,Rollins F,Kendall J,Edwards L,Singh VA,Stone GC,Schatz MC,Hicks J,Hannon GJ,Sordella R
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Mon Jan 16 00:00:00 EST 2017
Increased infiltration and tolerised antigen-specific CD8+ TEM cells in tumor but not peripheral blood have no impact on survival of HCMV+ glioblastoma patients.
Oncoimmunology
Bahador M,Gras Navarro A,Rahman MA,Dominguez-Valentin M,Sarowar S,Ulvestad E,Njølstad G,Lie SA,Kristoffersen EK,Bratland E,Chekenya M
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Targeted Disruption of TCF12 Reveals HEB as Essential in Human Mesodermal Specification and Hematopoiesis.
Stem cell reports
Li Y,Brauer PM,Singh J,Xhiku S,Yoganathan K,Zúñiga-Pflücker JC,Anderson MK
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Sep 12 00:00:00 EDT 2017
Region and cell-type resolved quantitative proteomic map of the human heart.
Nature communications
Doll S,Dreßen M,Geyer PE,Itzhak DN,Braun C,Doppler SA,Meier F,Deutsch MA,Lahm H,Lange R,Krane M,Mann M
25-0319 was used in Flow cytometry/Cell sorting to determine the healthy human heart proteome.
Mon Nov 13 00:00:00 EST 2017
CXCL12/CXCR4 activation by cancer-associated fibroblasts promotes integrin β1 clustering and invasiveness in gastric cancer.
International journal of cancer
Izumi D,Ishimoto T,Miyake K,Sugihara H,Eto K,Sawayama H,Yasuda T,Kiyozumi Y,Kaida T,Kurashige J,Imamura Y,Hiyoshi Y,Iwatsuki M,Iwagami S,Baba Y,Sakamoto Y,Miyamoto Y,Yoshida N,Watanabe M,Takamori H,Araki N,Tan P,Baba H
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2016
No Significant Reduction of Circulating Endothelial-Derived and Platelet-Derived Microparticles in Patients with Psoriasis Successfully Treated with Anti-IL12/23.
BioMed research international
Ho JC,Lee CH,Lin SH
25-0319 was used in Flow cytometry/Cell sorting to assess the impact of anti-IL-12/13 treatment on the development of cardiovascular disease.
Fri Feb 10 00:00:00 EST 2017
Notch ligand Delta-like 1 promotes in vivo vasculogenesis in human cord blood-derived endothelial colony forming cells.
Cytotherapy
Kim H,Huang L,Critser PJ,Yang Z,Chan RJ,Wang L,Carlesso N,Voytik-Harbin SL,Bernstein ID,Yoder MC
25-0319 was used in Flow cytometry/Cell sorting to conclude that Notch pathway activation in endothelial colony forming cells promotes vasculogenesis and augments blood vessel formation via diminishing apoptosis.
Fri May 01 00:00:00 EDT 2015
Activation of the Wnt/planar cell polarity pathway is required for pericyte recruitment during pulmonary angiogenesis.
The American journal of pathology
Yuan K,Orcholski ME,Panaroni C,Shuffle EM,Huang NF,Jiang X,Tian W,Vladar EK,Wang L,Nicolls MR,Wu JY,de Jesus Perez VA
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Thu Jan 01 00:00:00 EST 2015
Human esophageal myofibroblasts secrete proinflammatory cytokines in response to acid and Toll-like receptor 4 ligands.
American journal of physiology. Gastrointestinal and liver physiology
Gargus M,Niu C,Vallone JG,Binkley J,Rubin DC,Shaker A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Mon Jun 01 00:00:00 EDT 2015
Vascular progenitors from cord blood-derived induced pluripotent stem cells possess augmented capacity for regenerating ischemic retinal vasculature.
Circulation
Park TS,Bhutto I,Zimmerlin L,Huo JS,Nagaria P,Miller D,Rufaihah AJ,Talbot C,Aguilar J,Grebe R,Merges C,Reijo-Pera R,Feldman RA,Rassool F,Cooke J,Lutty G,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Jan 21 00:00:00 EST 2014
Hypoxia-cultured human adipose-derived mesenchymal stem cells are non-oncogenic and have enhanced viability, motility, and tropism to brain cancer.
Cell death & disease
Feng Y,Zhu M,Dangelmajer S,Lee YM,Wijesekera O,Castellanos CX,Denduluri A,Chaichana KL,Li Q,Zhang H,Levchenko A,Guerrero-Cazares H,Quiñones-Hinojosa A
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Thu Dec 11 00:00:00 EST 2014
Efficient and simultaneous generation of hematopoietic and vascular progenitors from human induced pluripotent stem cells.
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Park TS,Zimmerlin L,Zambidis ET
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Tue Jan 01 00:00:00 EST 2013
Efficient direct reprogramming of mature amniotic cells into endothelial cells by ETS factors and TGFβ suppression.
Cell
Ginsberg M,James D,Ding BS,Nolan D,Geng F,Butler JM,Schachterle W,Pulijaal VR,Mathew S,Chasen ST,Xiang J,Rosenwaks Z,Shido K,Elemento O,Rabbany SY,Rafii S
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Fri Oct 26 00:00:00 EDT 2012
ALDH1A isozymes are markers of human melanoma stem cells and potential therapeutic targets.
Stem cells (Dayton, Ohio)
Luo Y,Dallaglio K,Chen Y,Robinson WA,Robinson SE,McCarter MD,Wang J,Gonzalez R,Thompson DC,Norris DA,Roop DR,Vasiliou V,Fujita M
25-0319 was used in Flow cytometry/Cell sorting to demonstrate the existence of melanoma cells that fulfil the criteria for CSCs.
Mon Oct 01 00:00:00 EDT 2012
Cells expressing the C/EBPbeta isoform, LIP, engulf their neighbors.
PloS one
Abreu M,Sealy L
Published figure using CD31 (PECAM-1) monoclonal antibody (Product # 25-0319-42) in Flow Cytometry
Thu Apr 11 00:00:00 EDT 2013