Product References
Calcaratarin D, a labdane diterpenoid, attenuates mouse asthma via modulating alveolar macrophage function.
British journal of pharmacology
Liao W,Foo HYC,Tran TNQ,Chai CLL,Wong WSF
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2023
Nitric oxide controls proliferation of Leishmania major by inhibiting the recruitment of permissive host cells.
Immunity
Formaglio P,Alabdullah M,Siokis A,Handschuh J,Sauerland I,Fu Y,Krone A,Gintschel P,Stettin J,Heyde S,Mohr J,Philipsen L,Schröder A,Robert PA,Zhao G,Khailaie S,Dudeck A,Bertrand J,Späth GF,Kahlfuß S,Bousso P,Schraven B,Huehn J,Binder S,Meyer-Hermann M,Müller AJ
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Tue Dec 14 00:00:00 EST 2021
Prostaglandin E3 attenuates macrophage-associated inflammation and prostate tumour growth by modulating polarization.
Journal of cellular and molecular medicine
Cui J,Shan K,Yang Q,Qi Y,Qu H,Li J,Wang R,Jia L,Chen W,Feng N,Chen YQ
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Anisakis pegreffii Extract Induces Airway Inflammation with Airway Remodeling in a Murine Model System.
BioMed research international
Choi JH,Kim JY,Yi MH,Kim M,Yong TS
25-5920-82 was used in Flow Cytometry to confirm that A. pegreffii plays an essential role in causing asthma in mouse models and has the potential to cause similar effects in humans.
Wed Jan 12 00:00:00 EST 2022
Regulation of the long noncoding RNA XIST on the inflammatory polarization of microglia in cerebral infarction.
Experimental and therapeutic medicine
Zhang M,Yang JK,Ma J
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Wed Sep 01 00:00:00 EDT 2021
Exosomes from human umbilical cord mesenchymal stem cells attenuate the inflammation of severe steroid-resistant asthma by reshaping macrophage polarization.
Stem cell research & therapy
Dong B,Wang C,Zhang J,Zhang J,Gu Y,Guo X,Zuo X,Pan H,Hsu AC,Wang G,Wang F
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Wed Mar 24 00:00:00 EDT 2021
Alphavirus-Driven Interferon Gamma (IFNg) Expression Inhibits Tumor Growth in Orthotopic 4T1 Breast Cancer Model.
Vaccines
Trofimova O,Korotkaja K,Skrastina D,Jansons J,Spunde K,Isaguliants M,Zajakina A
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Wed Oct 27 00:00:00 EDT 2021
PD-L1 Improves Motor Function and Alleviates Neuropathic Pain in Male Mice After Spinal Cord Injury by Inhibiting MAPK Pathway.
Frontiers in immunology
Kong F,Sun K,Zhu J,Li F,Lin F,Sun X,Luo X,Ren C,Lu L,Zhao S,Sun J,Wang Y,Shi J
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Mon Nov 01 00:00:00 EDT 2021
TREM2 is a receptor for non-glycosylated mycolic acids of mycobacteria that limits anti-mycobacterial macrophage activation.
Nature communications
Iizasa E,Chuma Y,Uematsu T,Kubota M,Kawaguchi H,Umemura M,Toyonaga K,Kiyohara H,Yano I,Colonna M,Sugita M,Matsuzaki G,Yamasaki S,Yoshida H,Hara H
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri Apr 16 00:00:00 EDT 2021
Computational modeling reveals a key role for polarized myeloid cells in controlling osteoclast activity during bone injury repair.
Scientific reports
Lo CH,Baratchart E,Basanta D,Lynch CC
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Mon Mar 15 00:00:00 EDT 2021
Gasdermin D in peripheral nerves: the pyroptotic microenvironment inhibits nerve regeneration.
Cell death discovery
Tao Y,Wang F,Xu Z,Lu X,Yang Y,Wu J,Yao C,Yi F,Li J,Huang Z,Liu Y
25-5920-82 was used in Flow Cytometry to study the involvement of foamy cells in peripheral nerve regeneration in a GSDMD-/- mouse model.
Mon Jun 14 00:00:00 EDT 2021
Age-related exacerbation of hematopoietic organ damage induced by systemic hyper-inflammation in senescence-accelerated mice.
Scientific reports
Harada T,Tsuboi I,Hino H,Yuda M,Hirabayashi Y,Hirai S,Aizawa S
25-5920-82 was used in Flow cytometry/Cell sorting to find that the dosing of SAMP1/TA-1 mice with LPS revealed that the suppression of myelopoiesis and B-lymphopoiesis was more severe in aged mice than in young mice.
Wed Dec 01 00:00:00 EST 2021
Senolytic CAR T cells reverse senescence-associated pathologies.
Nature
Amor C,Feucht J,Leibold J,Ho YJ,Zhu C,Alonso-Curbelo D,Mansilla-Soto J,Boyer JA,Li X,Giavridis T,Kulick A,Houlihan S,Peerschke E,Friedman SL,Ponomarev V,Piersigilli A,Sadelain M,Lowe SW
25-5920 was used in Flow cytometry/Cell sorting to establish the therapeutic potential of senolytic CAR T cells for senescence-associated diseases.
Wed Jul 01 00:00:00 EDT 2020
Induced Tumor Heterogeneity Reveals Factors Informing Radiation and Immunotherapy Combinations.
Clinical cancer research : an official journal of the American Association for Cancer Research
Aguilera TA,Elghonaimy EA,Shehade H,Rafat M,Castellini L,Jiang D,Kariolis M,Koong AC,Le QT,Ellies LG,Rankin EB,Graves EE,Giaccia AJ
25-5920 was used in Flow cytometry/Cell sorting to investigate how induced tumor heterogeneity influences immune responses to radiotherapy with different proportions of mixed immune-responsive and unresponsive tumor cells in a triple-negative breast cancer model.
Mon Jun 15 00:00:00 EDT 2020
Modulation of the functions of myeloid-derived suppressor cells : a new strategy of hydrogen sulfide anti-cancer effects.
British journal of pharmacology
De Cicco P,Ercolano G,Rubino V,Terrazzano G,Ruggiero G,Cirino G,Ianaro A
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Sat Feb 01 00:00:00 EST 2020
ZBTB20-mediated titanium particle-induced peri-implant osteolysis by promoting macrophage inflammatory responses.
Biomaterials science
Qiu J,Peng P,Xin M,Wen Z,Chen Z,Lin S,Kuang M,Fu Y,Fang G,Li S,Li C,Mao J,Qin L,Ding Y
25-5920 was used in Flow cytometry/Cell sorting to study the molecular mechanism(s) of zinc finger and BTB domain-containing protein 20 (ZBTB20) contributing to titanium particle (TiP)-induced macrophage inflammatory responses and osteolysis.
Sun Jun 07 00:00:00 EDT 2020
Anticancer Activity of Liquid Treated with Microwave Plasma-Generated Gas through Macrophage Activation.
Oxidative medicine and cellular longevity
Lee CB,Seo IH,Chae MW,Park JW,Choi EH,Uhm HS,Baik KY
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Mycobacterium tuberculosis-infected alveolar epithelial cells modulate dendritic cell function through the HIF-1α-NOS2 axis.
Journal of leukocyte biology
Rodrigues TS,Alvarez ARP,Gembre AF,Forni MFPAD,de Melo BMS,Alves Filho JCF,Câmara NOS,Bonato VLD
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2020
Enhanced alleviation of aGVHD by TGF-β1-modified mesenchymal stem cells in mice through shifting MΦ into M2 phenotype and promoting the differentiation of Treg cells.
Journal of cellular and molecular medicine
Wu R,Liu C,Deng X,Chen L,Hao S,Ma L
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Wed Jan 01 00:00:00 EST 2020
Mycobacterium tuberculosis Heat-Shock Protein 16.3 Induces Macrophage M2 Polarization Through CCRL2/CX3CR1.
Inflammation
Zhang Y,Li S,Liu Q,Long R,Feng J,Qin H,Li M,Liu L,Luo J
25-5920 was used in Flow cytometry/Cell sorting to provides evidence that Mycobacterium tuberculosis (MTB) Hsp16.3 promotes macrophages to M2 phenotype and explores its underlying mechanism.
Wed Apr 01 00:00:00 EDT 2020
Proimmunogenic impact of MEK inhibition synergizes with agonist anti-CD40 immunostimulatory antibodies in tumor therapy.
Nature communications
Baumann D,Hägele T,Mochayedi J,Drebant J,Vent C,Blobner S,Noll JH,Nickel I,Schumacher C,Boos SL,Daniel AS,Wendler S,Volkmar M,Strobel O,Offringa R
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri May 01 00:00:00 EDT 2020
CCL2 and CXCL12 Derived from Mesenchymal Stromal Cells Cooperatively Polarize IL-10+ Tissue Macrophages to Mitigate Gut Injury.
Cell reports
Giri J,Das R,Nylen E,Chinnadurai R,Galipeau J
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Tue Feb 11 00:00:00 EST 2020
Elevated granulocytic myeloid-derived suppressor cells are closely related with elevation of Th17 cells in mice with experimental asthma.
International journal of biological sciences
Xue F,Yu M,Li L,Zhang W,Ma Y,Dong L,Shan W,Zheng Y,Wang T,Feng D,Lv J,Wang X
25-5920 was used in Flow cytometry/Cell sorting to indicate that G-MDSC elevation plays a crucial role in asthmatic mice.
Fri Nov 12 00:00:00 EST 2021
Inducing cancer indolence by targeting mitochondrial Complex I is potentiated by blocking macrophage-mediated adaptive responses.
Nature communications
Kurelac I,Iommarini L,Vatrinet R,Amato LB,De Luise M,Leone G,Girolimetti G,Umesh Ganesh N,Bridgeman VL,Ombrato L,Columbaro M,Ragazzi M,Gibellini L,Sollazzo M,Feichtinger RG,Vidali S,Baldassarre M,Foriel S,Vidone M,Cossarizza A,Grifoni D,Kofler B,Malanchi I,Porcelli AM,Gasparre G
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri Feb 22 00:00:00 EST 2019
Platelets Promote Macrophage Polarization toward Pro-inflammatory Phenotype and Increase Survival of Septic Mice.
Cell reports
Carestia A,Mena HA,Olexen CM,Ortiz Wilczyñski JM,Negrotto S,Errasti AE,Gómez RM,Jenne CN,Carrera Silva EA,Schattner M
25-5920 was used in Flow cytometry/Cell sorting to show the clinical outcome of sepsis is improved through by platelets that affect macrophage effector responses.
Tue Jul 23 00:00:00 EDT 2019
Adoptive transfer of Trichinella spiralis-activated macrophages can ameliorate both Th1- and Th2-activated inflammation in murine models.
Scientific reports
Kang SA,Park MK,Park SK,Choi JH,Lee DI,Song SM,Yu HS
25-5920-82 was used in Flow cytometry/Cell sorting to compare the effects of M1 and M2 macrophages obtained from Trichinella spiralis-infected mice with those of T. spiralis excretory/secretory (ES) protein-treated macrophages on experimental intestinal inflammation and allergic airway inflammation.
Thu Apr 25 00:00:00 EDT 2019
Chloroquine modulates antitumor immune response by resetting tumor-associated macrophages toward M1 phenotype.
Nature communications
Chen D,Xie J,Fiskesund R,Dong W,Liang X,Lv J,Jin X,Liu J,Mo S,Zhang T,Cheng F,Zhou Y,Zhang H,Tang K,Ma J,Liu Y,Huang B
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Wed Feb 28 00:00:00 EST 2018
Oncostatin M is overexpressed in skin squamous-cell carcinoma and promotes tumor progression.
Oncotarget
Simonneau M,Frouin E,Huguier V,Jermidi C,Jégou JF,Godet J,Barra A,Paris I,Levillain P,Cordier-Dirikoc S,Pedretti N,Bernard FX,Lecron JC,Morel F,Favot L
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri Nov 23 00:00:00 EST 2018
Mitochondrial permeabilization engages NF-κB-dependent anti-tumour activity under caspase deficiency.
Nature cell biology
Giampazolias E,Zunino B,Dhayade S,Bock F,Cloix C,Cao K,Roca A,Lopez J,Ichim G,Proïcs E,Rubio-Patiño C,Fort L,Yatim N,Woodham E,Orozco S,Taraborrelli L,Peltzer N,Lecis D,Machesky L,Walczak H,Albert ML,Milling S,Oberst A,Ricci JE,Ryan KM,Blyth K,Tait SWG
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri Sep 01 00:00:00 EDT 2017
Depletion of microglia exacerbates postischemic inflammation and brain injury.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism
Jin WN,Shi SX,Li Z,Li M,Wood K,Gonzales RJ,Liu Q
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
TGF-β receptor 1 inhibition prevents stenosis of tissue-engineered vascular grafts by reducing host mononuclear phagocyte activation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Lee YU,de Dios Ruiz-Rosado J,Mahler N,Best CA,Tara S,Yi T,Shoji T,Sugiura T,Lee AY,Robledo-Avila F,Hibino N,Pober JS,Shinoka T,Partida-Sanchez S,Breuer CK
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Fri Jul 01 00:00:00 EDT 2016
TNF-Mediated Restriction of Arginase 1 Expression in Myeloid Cells Triggers Type 2 NO Synthase Activity at the Site of Infection.
Cell reports
Schleicher U,Paduch K,Debus A,Obermeyer S,König T,Kling JC,Ribechini E,Dudziak D,Mougiakakos D,Murray PJ,Ostuni R,Körner H,Bogdan C
Published figure using iNOS monoclonal antibody (Product # 25-5920-82) in Flow Cytometry
Tue May 03 00:00:00 EDT 2016
Liver Necrosis and Lethal Systemic Inflammation in a Murine Model of Rickettsia typhi Infection: Role of Neutrophils, Macrophages and NK Cells.
PLoS neglected tropical diseases
Papp S,Moderzynski K,Rauch J,Heine L,Kuehl S,Richardt U,Mueller H,Fleischer B,Osterloh A
25592082 was used in flow cytometry to investigate the contribution of neutrophils and macrophages during Rickettsia typhi infection
Mon Aug 01 00:00:00 EDT 2016
Commensal Gram-positive bacteria initiates colitis by inducing monocyte/macrophage mobilization.
Mucosal immunology
Nakanishi Y,Sato T,Ohteki T
Thu Jan 01 00:00:00 EST 2015
Maternal immune activation evoked by polyinosinic:polycytidylic acid does not evoke microglial cell activation in the embryo.
Frontiers in cellular neuroscience
Smolders S,Smolders SM,Swinnen N,Gärtner A,Rigo JM,Legendre P,Brône B
25-5920 was used in Flow cytometry/Cell sorting to indicate that activation of the fetal microglial cells is not likely to be responsible for the inflammation induced deficits in this model for neurodevelopmental disorders.
Mon Aug 24 00:00:00 EDT 2015