    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ZONAB Antibodies
          Invitrogen
          ZONAB Polyclonal Antibody
          21 Published Figures
          23 References
          View all (14) ZONAB antibodies
          Questions & Answers
          Cite ZONAB Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Published Figures (21)
          ZONAB Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          ZONAB Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          FIGURE: 1 / 23
          ZONAB Antibody (40-2800) in IHC
          Indirect immunofluorescent staining of mouse liver using our ZONAB (Mid) Polyclonal Antibody, Rabbit (Product # 40-2800). Image courtesy of James I. Nagy, PhD, University of Manitoba, Canada {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Western Blot (WB)
          ZONAB Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZONAB Antibody in Immunocytochemistry (ICC/IF)
          ZONAB Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZONAB Antibody in Immunocytochemistry (ICC/IF)
          ZONAB Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZONAB Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Immunohistochemistry (IHC)
          ZONAB Antibody in Western Blot (WB)
          ZONAB Antibody in Western Blot (WB)
          ZONAB Antibody in Western Blot (WB)
          ZONAB Antibody in Western Blot (WB)
          ZONAB Antibody in Immunomicroscopy (IM)
          Product Details
          40-2800
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          3-4 µg/mL
          View 10 publications 10 publications
          Immunohistochemistry (IHC)
          -
          View 6 publications 6 publications
          Immunohistochemistry (Frozen) (IHC (F))
          2-3 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          -
          View 4 publications 4 publications
          Immunoprecipitation (IP)
          1-2 µg
          View 1 publication 1 publication
          Immunomicroscopy (IM)
          -
          View 1 publication 1 publication
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Published species
          Dog, Human, Mouse, Rabbit, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthetic peptide derived from an internal region of mouse ZONAB (ZO-1 associated nucleic acid-binding protein
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.25 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4
          Contains
          0.1% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2533460
          Product Specific Information
          This antibody reacts with an internal region of the ZONAB protein. On Western blots, it identifies a major band at ~65 kDa and a minor band at ~54 kDa. The antibody is directed towards a region of ZONAB common to both the long and the alternatively-spliced short form. During development, reactivity was confirmed using mouse testis tissue homogenates by Western blotting and labeling of mouse testis tissue using immunofluorescence. All signals were absent in knockout mice, further confirming the reactivity with ZONAB.
          Target Information
          Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
          Bioinformatics
          Protein Aliases: cold shock domain protein A; Cold shock domain-containing protein A; DNA-binding protein A; Muscle Y-box protein YB2; RYB-A; Y-box protein 3; Y-box-binding protein 3; Y-box-binding protein A; ZONAB A; ZONAB B
          View more View less
          Gene Aliases: Csda; Dbpa; Dpba; MSY3; MSY4; oxyR; Yb2; Ybx3
          View more View less
          UniProt ID: (Rat) Q62764, (Mouse) Q80WG4
          View more View less
          Entrez Gene ID: (Rat) 83807, (Mouse) 56449
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding single-stranded DNA binding mRNA 3'-UTR binding poly(A) RNA binding nucleic acid binding RNA binding protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter in utero embryonic development transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter spermatogenesis male gonad development fertilization organ regeneration negative regulation of apoptotic process positive regulation of organ growth negative regulation of skeletal muscle tissue development negative regulation of necroptotic process cellular response to tumor necrosis factor cellular hyperosmotic response negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline