    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DEFB104A Antibodies
          Invitrogen
          DEFB104A Polyclonal Antibody, PeproTech®
          View all (4) DEFB104A antibodies
          Tech Support
          Cite DEFB104A Polyclonal Antibody, PeproTech®
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          DEFB104A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DEFB104A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          DEFB104A Antibody (500-P268-100UG) in WB
          Western Blot: To detect Human BD-4 by Western Blot analysis DEFB104A Polyclonal Antibody (Product # 500-P268-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DEFB104A Antibody in Western Blot (WB)
          DEFB104A Antibody in Western Blot (WB)
          DEFB104A Antibody in ELISA (ELISA)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          DEFB104A Polyclonal Antibody, PeproTech®
          Product Details
          500-P268-100UG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.1-0.2 µg/mL
          -
          ELISA (ELISA)
          0.5-2.0 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          E.coli-derived Recombinant Human BD-4
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.1-1.0 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          RRID
          AB_2930158
          Product Specific Information
          AA Sequence of recombinant protein: EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP.
          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-4. Anti-Human BD-4-specific antibody was purified by affinity chromatography employing an immobilized Human BD-4 matrix.
          Sandwich ELISA: To detect Human BD-4 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-4 (500-P268Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-4.
          Western Blot: To detect Human BD-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BD-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
          Target Information
          Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.
          Bioinformatics
          Protein Aliases: defensin, beta 4
          View more View less
          Gene Aliases: BD-4; DEFB-4; DEFB104; DEFB4; hBD-4
          View more View less
          UniProt ID: (Human) Q8WTQ1
          View more View less
          Entrez Gene ID: (Human) 140596
          View more View less
          Function(s)
          chemoattractant activity
          Process(es)
          monocyte chemotaxis innate immune response defense response to Gram-negative bacterium defense response to Gram-positive bacterium positive chemotaxis cellular response to phorbol 13-acetate 12-myristate
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-wtp66:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0