Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HTR7 Antibodies

          Osenses

          5HT7 R Polyclonal Antibody

          1 Reference
          View all (18) HTR7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite 5HT7 R Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          5HT7 R Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          5HT7 R Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          5HT7 R Antibody (OSS00205W-100UL) in IHC (P)

          IHC-P on paraffin sections of rat brain. The animal was perfused using Autoperfuser at a pressure of 130 mmHg with 300 mL 4% FA being processed for paraffin embedding. HIER: Tris-EDTA, pH 9 for 20 min using Thermo PT Module. Blocking: 0.2% LFDM in TBST filtered thru 0.2 µm. Detection was done using Novolink HRP polymer from Leica following manufacturers instructions; DAB chromogen: Candela DAB chromogen fro... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          5HT7 R Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          5HT7 R Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          OSS00205W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:1,000
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Guinea pig, Human, Mouse, Rat

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from human 5HT7 R conjugated to blue carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in rat, mouse and guinea pig.

          Specificity of this antibody: 5HT7R receptor.

          Target Information

          5-HT7 Receptor is a Serotonin Receptor with several functions, including regulation of the circadian rhythm and mediation of smooth-muscle relaxation in the vasculature and GI tract. This receptor has been implicated in depression, autism, and other neuropsychiatric disorders. Three protein isoforms have been documented: isoform a: 445aa; isoform b: 432aa; isoform d: 479aa. 5-HT7 Receptor has been reported in aorta, brain, heart, placenta, small intestine, spleen, artery, and vessel. An EST has been isolated from a testis library.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 5-HT-7; 5-HT-X; 5-HT7; 5-HT7 receptor; 5-HT7b receptor; G protein coupled receptor; 5-HT7c receptor; G protein coupled receptor; 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled); 5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled; 5-hydroxytryptamine7 receptor; 5HT7 Receptor; GPRFO; high affinity serotonin receptor (5HT7); HT7; HTR7B; G-protein coupled receptor; OTTHUMP00000020077; OTTHUMP00000020078; OTTHUMP00000020079; serotonin receptor 7; unnamed protein product

          View more View less

          Gene Aliases: 5-HT-X; 5-HT7; 5Ht7

          View more View less

          UniProt ID: (Human) P34969, (Rat) P32305, (Guinea pig) P50407, (Mouse) P32304

          View more View less

          Entrez Gene ID: (Human) 3363, (Rat) 65032, (Guinea pig) 100135549, (Mouse) 15566

          View more View less

          Function(s)
          G-protein coupled receptor activity G-protein coupled serotonin receptor activity protein binding neurotransmitter receptor activity serotonin receptor activity G-protein coupled receptor
          Process(es)
          smooth muscle contraction G-protein coupled receptor signaling pathway G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger adenylate cyclase-activating serotonin receptor signaling pathway chemical synaptic transmission circadian rhythm blood circulation vasoconstriction G protein-coupled serotonin receptor signaling pathway serotonin receptor signaling pathway memory positive regulation of neuron projection development negative regulation of serotonin secretion urinary bladder smooth muscle contraction behavioral response to pain detection of mechanical stimulus involved in sensory perception of pain response to corticosterone response to electrical stimulus long-term synaptic potentiation regulation of vein smooth muscle contraction dopaminergic neuron differentiation presynaptic modulation of chemical synaptic transmission
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0