    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHMP7 Antibodies
          Proteintech
          CHMP7 Monoclonal Antibody (2F8F6)
          View all (12) CHMP7 antibodies
          Questions & Answers
          Cite CHMP7 Monoclonal Antibody (2F8F6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          CHMP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CHMP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          CHMP7 Antibody (68406-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed A431 cells using CHMP7 antibody (68406-1-Ig, Clone: 2F8F6) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CHMP7 Antibody in Immunocytochemistry (ICC/IF)
          CHMP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHMP7 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CHMP7 Antibody in Western Blot (WB)
          CHMP7 Monoclonal Antibody (2F8F6)
          Product Details
          68406-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:1,000-1:4,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2F8F6
          Immunogen
          CHMP7 fusion protein Ag9641
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: ELQRESDFMA SVDSSWISWG VGVFLLKPLK WTLSNMLGDN KVPAEEVLVA VELLKEKAEE VYRLYQNSPL SSHPVVALSE LSTLCANSCP DERTFYLVLL QLQKEKRVTV LEQNGEKIVK FARGPRAKVS PVNDVDVGVY QLMQSEQLLS RKVESLSQEA ERCKEEARRA CRAGKKQLAL RSLKAKQRTE KRIEALHAKL DTVQGILDRI YASQTDQMVF NAYQAGVGAL KLSMKDVTVE KAESLVDQIQ ELCDTQDEVS QTLAGGVTNG LDFDSEELEK ELDILLQDTT KEPLDLPDNP RNRHFTNSVP NPRISDAELE AELEKLSLSE GGLVPSSKSP KRQLEPTLKP L
          Target Information
          CHMP7 gene ontology annotations related to this gene include vacuolar transport.
          Bioinformatics
          Protein Aliases: Charged multivesicular body protein 7; CHMP family, member 7; Chromatin-modifying protein 7; MGC29816; unnamed protein product
          View more View less
          Gene Aliases: CHMP7; RGD1308779
          View more View less
          UniProt ID: (Human) Q8WUX9
          View more View less
          Entrez Gene ID: (Human) 91782, (Rat) 364419
          View more View less
          Function(s)
          protein binding protein transporter activity membrane traffic protein
          Process(es)
          plasma membrane repair membrane budding autophagy nucleus organization vacuolar transport mitotic metaphase plate congression exit from mitosis protein transport nuclear envelope reassembly late endosome to vacuole transport via multivesicular body sorting pathway multivesicular body assembly viral budding via host ESCRT complex ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway late endosome to vacuole transport viral budding from plasma membrane vesicle fusion with vacuole multivesicular body-lysosome fusion midbody abscission protein localization to chromatin multivesicular body sorting pathway membrane fission autophagosome maturation regulation of mitotic spindle assembly late endosome to lysosome transport ESCRT III complex disassembly cell separation after cytokinesis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline