    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TSH beta Antibodies
          NeoBiotechnologies
          Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Recombinant Rabbit Monoclonal Antibody (TSHb/7001R)
          View all (32) TSH beta antibodies
          Questions & Answers
          Cite Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Recombinant Rabbit Monoclonal Antibody (TSHb/7001R)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Antibody (7252-RBM3-P1) in SDS-PAGE
          SDS-PAGE analysis Purified TSH beta Rabbit Monoclonal Antibody (TSHb/7001R). Confirmation of Purity and Integrity of Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Thyroid Stimulating Hormone, beta (TSH beta) (Pituitary Marker) Antibody in SDS-PAGE (SDS-PAGE)
          Product Details
          7252-RBM3-P1
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1-2 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG, kappa
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          TSHb/7001R
          Immunogen
          Recombinant human TSH beta fragment
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
          Bioinformatics
          Protein Aliases: thyroid stimulating hormone beta; Thyroid-stimulating hormone subunit beta; Thyrotropin beta chain; Thyrotropin subunit beta; TSH; TSH-B
          View more View less
          Gene Aliases: TSH-B; TSH-BETA; TSHB
          View more View less
          UniProt ID: (Human) P01222
          View more View less
          Entrez Gene ID: (Human) 7252
          View more View less
          Function(s)
          receptor binding hormone activity
          Process(es)
          signal transduction G-protein coupled receptor signaling pathway cell-cell signaling anatomical structure morphogenesis
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline