    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UNCX Antibodies
          Antibodies Inc
          Uncx Monoclonal Antibody (N155/9), FL594
          Greener Choice
          View all (11) UNCX antibodies
          Questions & Answers
          Cite Uncx Monoclonal Antibody (N155/9), FL594
          Additional Information:
          {{banner.description}}
          Uncx Monoclonal Antibody (N155/9), FL594
          Product Details
          75-205-FL594
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N155/9
          Immunogen
          Fusion protein amino acids 191-530 (C-terminus) of mouse Uncx (accession number O75164) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Transcription factor involved in somitogenesis and neurogenesis. Required for the maintenance and differentiation of particular elements of the axial skeleton. May act upstream of PAX9. Plays a role in controlling the development of connections of hypothalamic neurons to pituitary elements, allowing central neurons to reach the peripheral blood circulation and to deliver hormones for control of peripheral functions.
          Bioinformatics
          Protein Aliases: Homeobox protein unc-4 homolog; Homeobox protein Uncx4.1; Unc4.1 homeobox; UNCX41
          View more View less
          Gene Aliases: Chx4; Uncx; Uncx4.1
          View more View less
          UniProt ID: (Mouse) O08934
          View more View less
          Entrez Gene ID: (Mouse) 22255
          View more View less
          Function(s)
          DNA binding sequence-specific DNA binding homeodomain transcription factor
          Process(es)
          cartilage condensation transcription, DNA-templated regulation of transcription, DNA-templated multicellular organism development pattern specification process nervous system development regulation of gene expression dorsal spinal cord development olfactory bulb interneuron differentiation cell differentiation common myeloid progenitor cell proliferation regulation of cell differentiation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline