    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • OS9 Antibodies
          Proteintech
          OS9 Recombinant Rabbit Monoclonal Antibody (230149B4)
          View all (14) OS9 antibodies
          Questions & Answers
          Cite OS9 Recombinant Rabbit Monoclonal Antibody (230149B4)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          OS9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          OS9 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          OS9 Antibody (82932-1-RR) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with 82932-1-RR (OS9 antibody) at dilution of 1:5,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          OS9 Antibody in Western Blot (WB)
          OS9 Antibody in Flow Cytometry (Flow)
          OS9 Antibody in Surface plasmon resonance (SPR)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          OS9 Recombinant Rabbit Monoclonal Antibody (230149B4)
          Product Details
          82932-1-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230149B4
          Immunogen
          OS9 fusion protein Ag0106
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 750
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: GRHIQQYHME DSEIKGEVLY LGYYQSAFDW DDETAKASKQ HRLKRYHSQT YGNGSKCDLN GRPREAEVRF LCDEGAGISG DYIDRVDEPL SCSYVLTIRT PRLCPHPLLR PPPSAAPQAI LCHPSLQPEE YMAYVQRQAD SKQYGDKIIE ELQDLGPQVW SETKSGVAPQ KMAGASPTKD DSKDSDFWKM LNEPEDQAPG GEEVPAEEQD PSPEAADSAS GAPNDFQNNV QVKVIRSPAD LIRFIEELKG GTKKGKPNIG QEQPVDDAAE VPQREPEKER GDPERQREME EEEDEDEDED EDEDERQLLG E
          Target Information
          OS9 is an ER glycoprotein found in the intralumenal level and highly expressed in tumor tissue. The OS9 gene is transcriptionally induced upon activation of the Ire1/Xbp1 ER-stress pathway.
          Bioinformatics
          Protein Aliases: amplified in osteosarcoma 9; endoplasmic reticulum lectin 2; erlectin 2; OS-9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; unnamed protein product
          View more View less
          Gene Aliases: ERLEC2; OS-9
          View more View less
          UniProt ID: (Human) Q13438
          View more View less
          Entrez Gene ID: (Human) 10956
          View more View less
          Function(s)
          protease binding protein binding glycosylation-dependent protein binding
          Process(es)
          ubiquitin-dependent protein catabolic process protein targeting protein retention in ER lumen protein ubiquitination endoplasmic reticulum unfolded protein response retrograde protein transport, ER to cytosol response to endoplasmic reticulum stress ERAD pathway negative regulation of retrograde protein transport, ER to cytosol
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch