    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DDX39 Antibodies
          Proteintech
          DDX39A Recombinant Rabbit Monoclonal Antibody (230375F9)
          View all (8) DDX39 antibodies
          Questions & Answers
          Cite DDX39A Recombinant Rabbit Monoclonal Antibody (230375F9)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          DDX39A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DDX39A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          DDX39A Antibody (83083-5-RR) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HepG2 cells using DDX39 antibody (83083-5-RR, Clone: 230375F9) at dilution of 1:400 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG (H+L) (SA00013-1), CL594-Phalloidin (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DDX39A Antibody in Immunocytochemistry (ICC/IF)
          DDX39A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          DDX39A Antibody in Western Blot (WB)
          DDX39A Antibody in Western Blot (WB)
          DDX39A Antibody in Western Blot (WB)
          DDX39A Antibody in Flow Cytometry (Flow)
          DDX39A Antibody in Surface plasmon resonance (SPR)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          DDX39A Recombinant Rabbit Monoclonal Antibody (230375F9)
          Product Details
          83083-5-RR
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.25 µg/10^6 cells in a 100 µL suspension
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Surface plasmon resonance (SPR)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230375F9
          Immunogen
          DDX39A fusion protein Ag2311
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAEQDVENDL LDYDEEEEPQ APQESTPAPP KKDIKGSYVS IHSSGFRDFL LKPELLRAIV DCGFEHPSEV QHECIPQAIL GMDVLCQAKS GMGKTAVFVL ATLQQIEPVN GQVTVLVMCH TRELAFQISK EYERFSKYMP SVKVSVFFGG LSIKKDEEVL KKNCPHVVVG TPGRILALVR NRSFSLKNVK HFVLDECDKM LEQLDMRRDV QEIFRLTPHE KQCMMFSATL SKDIRPVCRK FMQDPMEVFV DDETKLTLHG LQQYYVKLKD SEKNRKLFDL LDVLEFNQPV TLSAVQGFPA ADPGGHQSVW PGDGHRASQH RL
          Target Information
          This gene encodes a member of the DEAD box protein family. These proteins are characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD) and are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of the DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene is thought to play a role in the prognosis of patients with gastrointestinal stromal tumors. A pseudogene of this gene is present on chromosome 13. Alternate splicing results in multiple transcript variants. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.
          Bioinformatics
          Protein Aliases: ATP-dependent RNA helicase DDX39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39; DEAD (Asp-Glu-Ala-Asp) box polypeptide 39A; DEAD box protein 39; DEAD-box helicase 39A; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 39; nuclear RNA helicase URH49; nuclear RNA helicase, DECD variant of DEAD box family; UAP56-related helicase, 49 kDa; unnamed protein product
          View more View less
          Gene Aliases: 2610307C23Rik; BAT1; BAT1L; DDX39; DDXL; URH49
          View more View less
          UniProt ID: (Human) O00148, (Mouse) Q8VDW0
          View more View less
          Entrez Gene ID: (Human) 10212, (Mouse) 68278
          View more View less
          Function(s)
          nucleic acid binding RNA binding RNA helicase activity mRNA binding protein binding ATP binding ATPase activity identical protein binding
          Process(es)
          mRNA splicing, via spliceosome toll-like receptor signaling pathway cytoplasmic pattern recognition receptor signaling pathway mRNA export from nucleus negative regulation of innate immune response negative regulation of RNA export from nucleus RNA export from nucleus
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0