    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Phosphatidylinositol 3,4,5-trisphosphate Antibodies
          Invitrogen
          Phosphatidylinositol 3,4,5-trisphosphate Monoclonal Antibody (RC6F8)
          1 Published Figure
          10 References
          Questions & Answers
          Cite Phosphatidylinositol 3,4,5-trisphosphate Monoclonal Antibody (RC6F8)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          Phosphatidylinositol 3,4,5-trisphosphate Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phosphatidylinositol 3,4,5-trisphosphate Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Phosphatidylinositol 3,4,5-trisphosphate Antibody (A-21328) in ICC/IF
          Localization of phosphatidylinositol 3, 4, 5-triphosphate in unstimulated (left panel) and insulin-stimulated (right panel) 3T3-L1 cells detected using anti-phosphatidylinositol 3, 4, 5-triphosphate, mouse IgM monoclonal RC6F8 (Product # A-21328) and visualized using biotinylated goat anti-mouse IgM in conjunction with Alexa Fluor® 488 streptavidin (Product # S-11223). Image courtesy of Echelon Research Lab... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phosphatidylinositol 3,4,5-trisphosphate Antibody in Immunocytochemistry (ICC/IF)
          Phosphatidylinositol 3,4,5-trisphosphate Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          A-21328
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          View 8 publications 8 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Chemical, Mouse
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          RC6F8
          Immunogen
          phosphatidylinositol 3,4,5-diphosphate
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.2, with 0.1% BSA
          Contains
          5mM sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2535837
          Target Information
          Phosphatidylinositol (3, 4, 5)-trisphosphate (PtdIns(3, 4, 5)P3) commonly abbreviated to PIP3 is the product of the class I phosphoinositide 3-kinases (PI 3-kinases) activity on phosphatidylinositol (4, 5)-bisphosphate. PtdIns (3, 4, 5) P3 is dephophosphorylated by the phosphatase, PTEN on the 3 position and by SHIPs (SH2-containing inositol phosphatase) on the 5' position of the inositol ring. The PH domain in a number of proteins binds to PtdIns (3, 4, 5) P3. Such proteins include Akt/PKB, PDK1, Btk1 and ARNO. The generation of PtdIns (3, 4, 5) P3 at the plasma membrane upon the activation of class I PI 3-kinases causes these proteins to translocate to the plasma membrane and accordingly affects their activity. The PH domain allows binding between PtdIns (3, 4, 5) P3 and G-protein coupled receptor kinases (GRKs). This enhances the binding of the GRK to the plasma membrane. [National Center for Biotechnology Information. PubChem Compound Database; CID=53477782].
          Bioinformatics
          Protein Aliases: C12H24O22P4; phosphatidylinositol 3; phosphatidylinositol-3; PIP3
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline