    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Mouse IgM (Heavy chain) Secondary Antibodies
          Discover how to image with Invitrogen F(ab) secondary antibodies
          Invitrogen
          Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555
          4 Published Figures
          View all (10) Goat anti-Mouse IgM (Heavy chain) antibodies
          Questions & Answers
          Cite Goat anti-Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Published Figures (4)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody (A-21426) in ICC/IF
          Immunofluorescence analysis of Goat anti-Mouse IgM (Heavy Chain) Secondary Antibody, Alexa Fluor™ 555 (Product # A-21426) was performed using MCF 10A (positive model) and A431 (negative model) cells stained with Vimentin Monoclonal Antibody (Product # MA1-19168). The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, blocked with 1% BSA for 1 hour... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Western Blot (WB)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Mouse IgM (Heavy chain) Cross-Adsorbed Secondary Antibody in Immunohistochemistry (IHC)
          Product Details
          A-21426
          Applications
          Tested Dilution
          Western Blot (WB)
          1:10,000
          Immunohistochemistry (IHC)
          1-10 µg/mL
          Immunocytochemistry (ICC/IF)
          1-10 µg/mL
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Secondary Antibody
          Immunogen
          Mouse Mu immunoglobulin
          Conjugate
          Alexa Fluor™ 555 Alexa Fluor™ 555 Alexa Fluor™ 555
          • Alexa Fluor 350 
          • Alexa Fluor 488 
          • Alexa Fluor 546 
          • Alexa Fluor 568 
          • Alexa Fluor 594 
          • Alexa Fluor 633 
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          553/568 nm View spectra spectra
          Form
          Liquid
          Concentration
          2 mg/mL
          Amount
          500 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.5
          Contains
          5mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2535847
          Target
          IgM (Heavy chain)
          Cross Adsorption
          Against human IgG1, IgG2, IgG3, IgG4, IgA, human serum and purified human paraproteins prior to conjugation
          Antibody Form
          Whole Antibody
          Product Specific Information
          Product will be shipped at Room Temperature.
          Target Information
          Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody A-21426 on CiteAb
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline