    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Lucifer Yellow Antibodies
          Invitrogen
          Lucifer Yellow Polyclonal Antibody
          16 Published Figures
          79 References
          View all (2) Lucifer Yellow antibodies
          Questions & Answers
          Cite Lucifer Yellow Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Published Figures (16)
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 16
          Lucifer Yellow Antibody (A-5750) in ICC/IF
          eNeuro 2021 - Figure 7. TRPM4 contributes to spontaneous firing in SCN neurons. A , Rate histogram (10-s bin averages of instantaneous frequency) of spontaneous action potential firing (no TTX) in an SCN neuron recorded in cell-attached configuration, under control conditions and in the presence of 9-pt (30 mu m ). Example recordings from the indicated time points are provided (right). B , Photomicrograph of Lucifer yell... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Lucifer Yellow Antibody in Immunocytochemistry (ICC/IF)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Lucifer Yellow Antibody in Immunohistochemistry (IHC)
          Product Details
          A-5750
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          Assay-dependent
          View 57 publications 57 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          -
          View 1 publication 1 publication
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          -
          View 1 publication 1 publication
          Immunohistochemistry (Frozen) (IHC (F))
          -
          View 2 publications 2 publications
          Immunohistochemistry - Free Floating (IHC (Free))
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          View 16 publications 16 publications
          Miscellaneous PubMed (Misc)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Chemical
          Published species
          Chemical, Mouse
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Lucifer Yellow
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          3 mg/mL
          Amount
          1.5 mg
          Purification
          IgG fraction
          Storage buffer
          PBS, pH 7.2
          Contains
          5mM sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2536190
          Target Information
          Even though the cell soma of a lucifer yellow-filled neuron may be brightly stained, its finer processes can sometimes be faint and may fade rapidly or be obscured by the more intensely stained portions of the neuron. By using anti-lucifer yellow dye antibodies in conjunction with standard enzyme-mediated immunohistochemical methods one can develop a more permanent, fade-free signal for light microscopy.
          Bioinformatics
          Protein Aliases: Lucifer Yellow Dye
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline