    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Mouse IgG Secondary Antibodies
          Invitrogen
          Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Biotin
          View all (276) Goat anti-Mouse IgG antibodies
          Questions & Answers
          Cite Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          Mouse IgG (H+L) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Mouse IgG (H+L) Secondary Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Mouse IgG (H+L) Secondary Antibody (A28176) in WB
          Western blot analysis was performed on whole cell extracts (30 µg lysate) of K-562 (Lane 1) and PC-3 (Lane 2). The blots were probed with Anti-SOD1 Mouse Monoclonal Antibody (Product # MA1-105, 0.25 µg/mL) and detected by chemiluminescence using Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Biotin conjugate (Product # A28176) at dilutions 0.05 µg/mL (Fig. 1), 0.5 µg/mL (Fig. 2) and 1 µg/mL (Fig... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Mouse IgG (H+L) Secondary Antibody in Western Blot (WB)
          Mouse IgG (H+L) Secondary Antibody in Western Blot (WB)
          Mouse IgG (H+L) Secondary Antibody in Flow Cytometry (Flow)
          Mouse IgG (H+L) Secondary Antibody in ELISA (ELISA)
          Product Details
          A28176
          Applications
          Tested Dilution
          Western Blot (WB)
          0.05-1 µg/mL
          Flow Cytometry (Flow)
          2.5 µg/mL
          ELISA (ELISA)
          Assay-dependent
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Goat / IgG
          Expression System
          Expi293
          Class
          Recombinant Superclonal
          Type
          Secondary Antibody
          Immunogen
          Recombinant full-length protein
          Conjugate
          Biotin Biotin Biotin
          • Agarose
          • Alexa Fluor 488 
          • Alexa Fluor 488-10 nm colloidal gold
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 790 
          • Alexa Fluor Plus 405 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • Alexa Fluor Plus 800 
          • AP
          • bGal
          • DyLight 350
          • DyLight 405
          • DyLight 488
          • DyLight 550
          • DyLight 594
          • DyLight 633
          • DyLight 650
          • DyLight 680
          • DyLight 800 4X PEG
          • FITC
          • HRP
          • PE 
          • Rhodamine
          • TRITC
          • Unconjugated
          • WesternDot 625
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          1 mg
          Purification
          Gravity column chromatography
          Storage buffer
          PBS
          Contains
          0.1% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Wet ice
          RRID
          AB_2536162
          Target
          IgG
          Antibody Form
          Whole Antibody
          Product Specific Information
          The sensitivity and specificity of each lot is confirmed using ELISA.
          Minimal cross-reactivity with rabbit, rat, human, bovine, guinea pig and donkey IgG is observed.
          Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
          Target Information
          Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody A28176 on CiteAb
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline