    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SPEN Antibodies
          Bethyl Laboratories
          SHARP Polyclonal Antibody
          View all (4) SPEN antibodies
          Questions & Answers
          Cite SHARP Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SHARP Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          SHARP Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SHARP Antibody (A301-119A-T) in IP
          Detection of human SHARP by western blot of immunoprecipitates. Samples: Whole cell lysate (1.0 mg per IP reaction; 5% of IP loaded) from HEK293T cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-SHARP antibody (Product # A301-119A) (lot 3) used for IP at 6 µg per reaction. SHARP was also immunoprecipitated by a second antibody against a different epitope of SHARP (A301-120A). For blotting immunoprecipitated SHARP, A301-119A was used at 0.1 µg/mL. Detection: Chemiluminescence with an exposure time of 10 seco... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SHARP Antibody in Immunoprecipitation (IP)
          SHARP Polyclonal Antibody
          Product Details
          A301-119A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 1350 and 1400 of Human SMART/HDAC1 associated repressor protein (spen homolog, transcriptional regulator).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Based on 100% sequence identity, this antibody is predicted to react with Mouse
          Target Information
          May serve as a nuclear matrix platform that organizes and integrates transcriptional responses. In osteoblasts, supports transcription activation: synergizes with RUNX2 to enhance FGFR2-mediated activation of the osteocalcin FGF-responsive element (OCFRE). Has also been shown to be an essential corepressor protein, which probably regulates different key pathways such as the Notch pathway. Negative regulator of the Notch pathway via its interaction with RBPSUH, which prevents the association between NOTCH1 and RBPSUH, and therefore suppresses the transactivation activity of Notch signaling. Blocks the differentiation of precursor B-cells into marginal zone B-cells. Probably represses transcription via the recruitment of large complexes containing histone deacetylase proteins. May bind both to DNA and RNA.
          Bioinformatics
          Protein Aliases: Msx2 interacting nuclear target (MINT) homolog; Msx2-interacting protein; nuclear receptor transcription cofactor; SMART/HDAC1-associated repressor protein; SPEN homolog; spen homolog, transcriptional regulator; unnamed protein product
          View more View less
          Gene Aliases: HIAA0929; KIAA0929; MINT; RATARS; RBM15C; SHARP; SPEN
          View more View less
          UniProt ID: (Human) Q96T58
          View more View less
          Entrez Gene ID: (Human) 23013
          View more View less
          Function(s)
          nucleic acid binding DNA binding transcription corepressor activity RNA binding mRNA binding protein binding RNA polymerase II sequence-specific DNA binding transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter Notch signaling pathway chromatin silencing by small RNA negative regulation of transcription, DNA-templated positive regulation of neurogenesis random inactivation of X chromosome
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline