    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SLX4IP Antibodies
          Bethyl Laboratories
          SLX4IP/C20orf94 Polyclonal Antibody
          View all (4) SLX4IP antibodies
          Questions & Answers
          Cite SLX4IP/C20orf94 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SLX4IP/C20orf94 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          SLX4IP/C20orf94 Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SLX4IP/C20orf94 Antibody (A304-995A-T) in IP
          Detection of human SLX4IP/C20orf94 by western blot of immunoprecipitates. Samples: Whole cell lysate (0.5 or 1.0 mg per IP reaction; 20% of IP loaded) from HeLa cells prepared using NETN lysis buffer. Antibodies: Affinity purified rabbit anti-SLX4IP/C20orf94 antibody (Product # A304-995A) (lot 1) used for IP at 6 µg per reaction. SLX4IP/C20orf94 was also immunoprecipitated by rabbit anti-SLX4IP/C20orf94 ant... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SLX4IP/C20orf94 Antibody in Immunoprecipitation (IP)
          SLX4IP/C20orf94 Polyclonal Antibody
          Product Details
          A304-995A-T
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2-10 µg/mg lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Region between residue 175 to 225 of Human UPF0492 protein C20orf94.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          10 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          tris citrate/phosphate, pH 7-8
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          SLX4IP/C20orf94 interacts with SLX4/BTBD12.
          Bioinformatics
          Protein Aliases: Protein SLX4IP; SLX4-interacting protein
          View more View less
          Gene Aliases: bA204H22.1; bA254M13.1; C20orf94; dJ1099D15.3; SLX4IP
          View more View less
          UniProt ID: (Human) Q5VYV7
          View more View less
          Entrez Gene ID: (Human) 128710
          View more View less
          Function(s)
          protein binding
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline