Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ABI3BP Antibodies

          Abnova

          ABI3BP Polyclonal Antibody, MaxPab™

          View all (7) ABI3BP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ABI3BP Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ABI3BP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ABI3BP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ABI3BP Antibody (H00025890-B01P) in WB

          Western Blot analysis of ABI3BP expression in transfected 293T cell line by ABI3BP MaxPab polyclonal antibody. Lane 1: ABI3BP transfected lysate(118.25 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ABI3BP Antibody in Western Blot (WB)

          Product Details

          H00025890-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ABI3BP (NP_056244, 1 a.a. approximately 1075 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MRGGKCNMLS SLGCLLLCGS ITLALGNAQK LPKGKRPNLK VHINTTSDSI LLKFLRPSPN VKLEGLLLGY GSNVSPNQYF PLPAEGKFTE AIVDAEPKYL IVVRPAPPPS QKKSCSGKTR SRKPLQLVVG TLTPSSVFLS WGFLINPHHD WTLPSHCPND RFYTIRYREK DKEKKWIFQI CPATETIVEN LKPNTVYEFG VKDNVEGGIW SKIFNHKTVV GSKKVNGKIQ STYDQDHTVP AYVPRKLIPI TIIKQVIQNV THKDSAKSPE KAPLGGVILV HLIIPGLNET TVKLPASLMF EISDALKTQL AKNETLALPA ESKTPEVEKI SARPTTVTPE TVPRSTKPTT SSALDVSETT LASSEKPWIV PTAKISEDSK VLQPQTATYD VFSSPTTSDE PEISDSYTAT SDRILDSIPP KTSRTLEQPR ATLAPSETPF VPQKLEIFTS PEMQPTTPAP QQTTSIPSTP KRRPRPKPPR TKPERTTSAG TITPKISKSP EPTWTTPAPG KTQFISLKPK IPLSPEVTHT KPAPKQTPRA PPKPKTSPRP RIPQTQPVPK VPQRVTAKPK TSPSPEVSYT TPAPKDVLLP HKPYPEVSQS EPAPLETRGI PFIPMISPSP SQEELQTTLE ETDQSTQEPF TTKIPRTTEL AKTTQAPHRF YTTVRPRTSD KPHIRPGVKQ APRPSGADRN VSVDSTHPTK KPGTRRPPLP PRPTHPRRKP LPPNNVTGKP GSAGIISSGP ITTPPLRSTP RPTGTPLERI ETDIKQPTVP ASGEELENIT DFSSSPTRET DPLGKPRFKG PHVRYIQKPD NSPCSITDSV KRFPKEEATE GNATSPPQNP PTNLTVVTVE GCPSFVILDW EKPLNDTVTE YEVISRENGS FSGKNKSIQM TNQTFSTVEN LKPNTSYEFQ VKPKNPLGEG PVSNTVAFST ESADPRVSEP VSAGRDAIWT ERPFNSDSYS ECKGKQYVKR TWYKKFVGVQ LCNSLRYKIY LSDSLTGKFY NIGDQRGHGE DHCQFVDSFL DGRTGQQLTS DQLPIKEGYF RAVRQEPVQF GEIGGHTQIN YVQWYECGTT IPGKW

          Target Information

          ABI3BP is a protein coding gene. Gene ontology (GO) annotation include collagen-containing extracellular matrix; extracellular region; extracellular space; interstitial matrix.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ABI family, member 3 (NESH) binding protein; ABI gene family member 3-binding protein; ABI gene family, member 3 (NESH) binding protein; FLJ41743; FLJ41754; Nesh-binding protein; NeshBP; Target of Nesh-SH3; Tarsh; unnamed protein product

          View more View less

          Gene Aliases: ABI3BP; NESHBP; TARSH

          View more View less

          UniProt ID: (Human) Q7Z7G0

          View more View less

          Entrez Gene ID: (Human) 25890

          View more View less

          Function(s)
          extracellular matrix structural constituent
          Process(es)
          positive regulation of cell-substrate adhesion extracellular matrix organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0