Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This target displays homology in the following species: Cow: 85%; Dog: 86%; Guinea Pig: 92%; Horse: 83%; Human: 100%; Mouse: 100%; Rabbit: 91%; Rat: 100%
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene is highly expressed in testis and may be involved in human spermatogenesis. Alternative splicing results in multiple transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: a disintegrin and metalloprotease domain 29; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 29; ADAM; ADAM29-beta; ADAMs; cancer/testis antigen 73; epididymis secretory sperm binding protein; metallaproteinase-disintegrin (ADAM29); metalloendopeptidases; testis secretory sperm-binding protein Li 207a; unnamed protein product
Gene Aliases: CT73; svph1
UniProt ID: (Human) Q9UKF5
Entrez Gene ID: (Human) 11086, (Mouse) 244486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support