Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSSKKSKEK EKEKDKIKEK EKDSKDKEKD KKTVNGHTFS SIPVVGPISC SQCMKPFTNK DAYTCANCSA FVHKGCRESL ASCAKVKMKQ PKGSLQAHDT SSLPTVIMRN KPSQPKERPR SAVLLVDETA TTPIFANRRS QQSVSLSKSV SIQNITGVGN DENMSNTWKF LSHSTDSLNK ISKVNESTES LTDEGVGTDM NEGQLLGDFE IESKQLEAES WSRIIDSKFL KQQKKDVVKR QEVIYELMQT EFHHVRTLKI MSGVYSQGMM ADLLFEQQMV EKLFPCLDEL ISIHSQFFQR ILERKKESLV DKSEKNFLIK RIGDVLVNQF SGENAERLKK TYGKFCGQHN QSVNYFKDLY AKDKRFQAFV KKKMSSSVVR RLGIPECILL VTQRITKYPV LFQRILQCTK DNEVEQEDLA QSLSLVKDVI GAVDSKVASY EKKVRLNEIY TKTDSKSIMR MKSGQMFAKE DLKRKKLVRD GSVFLKNAAG RLKEVQAVLL TDILVFLQEK DQKYIFASLD QKSTVISLKK LIVREVAHEE KGLFLISMGM TDPEMVEVHA SSKEERNSWI QIIQDTINTL NRDEDEGIPS ENEEEKKMLD TRARELKEQL HQKDQKILLL LEEKEMIFRD MAECSTPLPE DCSPTHSPRV LFRSNTEEAL KGGPLMKSAI NEVEILQGLV SGNLGGTLGP TVSSPIEQDV VGPVSLPRRA ETFGGFDSHQ MNASKGGEKE EGDDGQDLRR TESDSGLKKG GNANLVFMLK RNSEQVVQSV VHLYELLSAL QGVVLQQDSY IEDQKLVLSE RALTRSLSRP SSLIEQEKQR SLEKQRQDLA NLQKQQAQYL EEKRRREREW EARERELRER EALLAQREEE VQQGQQDLEK EREELQQKKG TYQYDLERLR AAQKQLEREQ EQLRREAERL SQRQTERDLC QVSHPHTKLM RIPSFFPSPE EPPSPSAPSI AKSGSLDSEL SVSPKRNSIS RTHKDKGPFH ILSSTSQTNK GPEGQSQAPA STSASTRLFG LTKPKEKKEK KKKNKTSRSQ PGDGPASEVS AEGEEIFC
The A-kinase anchor proteins (AKAPs) are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A (PKA) and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternative splicing of this gene results in multiple transcript variants encoding different isoforms containing c-terminal dbl oncogene homology (DH) and pleckstrin homology (PH) domains. The DH domain is associated with guanine nucleotide exchange activation for the Rho/Rac family of small GTP binding proteins, resulting in the conversion of the inactive GTPase to the active form capable of transducing signals. The PH domain has multiple functions. Therefore, these isoforms function as scaffolding proteins to coordinate a Rho signaling pathway, function as protein kinase A-anchoring proteins and, in addition, enhance ligand-dependent activity of estrogen receptors alpha and beta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A kinase (PRKA) anchor protein 13; AKAP-Lbc; guanine nucleotide exchange factor; breast cancer nuclear receptor-binding auxiliary protein; Brx/Proto-Lbc; guanine nucleotide exchange factor Lbc; human thyroid-anchoring protein 31; LBC oncogene; lymphoid blast crisis oncogene; non-oncogenic Rho GTPase-specific GTP exchange factor; protein kinase A-anchoring protein 13; unnamed protein product
Gene Aliases: AKAP-13; AKAP-Lbc; ARHGEF13; BRX; c-lbc; HA-3; Ht31; LBC; p47; PRKA13; PROTO-LB; PROTO-LBC
UniProt ID: (Human) Q12802
Entrez Gene ID: (Human) 11214
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support