Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MQIIRHSEQ TLKTALISKN PVLVSQYEKL DAGEQRLMNE AFQPASDLFG PITLHSPSDW ITSHPEAPQD FEQFFSDPYR KTPSPNKRSI YIQSIGSLGN TRIISEEYIK WLTGYCKAYF YGLRVKLLEP VPVSVTRCSF RVNENTHNLQ IHAGDILKFL KKKKPEDAFC VVGITMIDLY PRDSWNFVFG QASLTDGVGI FSFARYGSDF YSMHYKGKVK KLKKTSSSDY SIFDNYYIPE ITSVLLLRSC KTLTHEIGHI FGLRHCQWLA CLMQGSNHLE EADRRPLNLC PICLHKLQCA VGFSIVERYK ALVRWIDDES SDTPGATPEH SHEDNGNLPK PVEAFKEWKE WIIKCLAVLQ K (1-360 aa encoded by BC056271)
AMZ2 (archaelysin family metallopeptidase 2), also known as archaemetzincin-2 or archeobacterial metalloproteinase-like protein 2, is a 360 amino acid protein belonging to the peptidase M54 family. Encoded by a gene that maps to human chromosome 17q24.2, AMZ2 is predominantly expressed in heart and testis. AMZ2 is also expressed in kidney, liver, pancreas, lung, brain and placenta, and in fetal tissues such as kidney, liver, lung and brain. AMZ2 participates in metal ion binding and functions as a zinc metalloprotease. AMZ2 is inhibited by both general metalloprotease inhibitors o-phenanthroline and batimastat. Exhibiting aminopeptidase activity, AMZ2 acts against Angiotensin in vitro, but does not hydrolyze either Neurogranin or Angiotensin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: archaemetzincins-2; archeobacterial metalloproteinase-like 2; archeobacterial metalloproteinase-like protein 2; unnamed protein product
Gene Aliases: ESTM12; RGD1304846
UniProt ID: (Human) Q86W34, (Mouse) Q400C8, (Rat) Q400C7
Entrez Gene ID: (Human) 51321, (Mouse) 13929, (Rat) 360650
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support