    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SLC28A3 Antibodies
          Alomone Labs, LTD
          SLC28A3/CNT3 (extracellular) Polyclonal Antibody
          View all (5) SLC28A3 antibodies
          Questions & Answers
          Cite SLC28A3/CNT3 (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          SLC28A3/CNT3 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SLC28A3/CNT3 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          SLC28A3/CNT3 (extracellular) Antibody (ANT-063-50UL) in WB
          Western blot analysis of human HL-60 myelocytic leukemia cell line lysate (lanes 1 and 5), human K562 chronic myelogenous leukemia cell line lysate (lanes 2 and 6), human MCF-7 breast adenocarcinoma cell line lysate (lanes 3 and 7) and human PANC-1 pancreatic carcinoma cell line lysate (lanes 4 and 8): - 1-4. Anti-SLC28A3/CNT3 (extracellular) Antibody (#ANT-063), (1:500). 5-8. Anti-SLC28A3/CNT3 (extracellul... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SLC28A3/CNT3 (extracellular) Antibody in Western Blot (WB)
          SLC28A3/CNT3 (extracellular) Antibody in Western Blot (WB)
          SLC28A3/CNT3 (extracellular) Antibody in Flow Cytometry (Flow)
          SLC28A3/CNT3 (extracellular) Polyclonal Antibody
          Product Details
          ANT-063-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:400
          -
          Flow Cytometry (Flow)
          5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Peptide (C)KITLKNAMKMENGDSR, corresponding to amino acid residues 432-447 of rat SLC28A3/CNT3, 5th extracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Bioinformatics
          Protein Aliases: broad substrate specificity for purines and pyrimidines (system cib); CNT 3; Concentrative Na(+)-nucleoside cotransporter 3; concentrative Na+-nucleoside cotransporter; mCNT3; rCNT3; solute carrier family 28 (concentrative nucleoside transporter), member 3; solute carrier family 28 (sodium-coupled nucleoside transporter), member 3; Solute carrier family 28 member 3; unnamed protein product
          View more View less
          Gene Aliases: CNT3; SLC28A3
          View more View less
          UniProt ID: (Human) Q9HAS3, (Rat) Q8VIH3, (Mouse) Q9ERH8
          View more View less
          Entrez Gene ID: (Human) 64078, (Rat) 140944, (Mouse) 114304
          View more View less
          Function(s)
          nucleoside transmembrane transporter activity purine nucleobase transmembrane transporter activity nucleoside:sodium symporter activity protein binding uridine transmembrane transporter activity symporter activity pyrimidine- and adenine-specific:sodium symporter activity purine-specific nucleoside:sodium symporter activity nucleoside binding transporter
          Process(es)
          xenobiotic metabolic process drug transmembrane transport pyrimidine nucleobase transport purine nucleoside transmembrane transport uridine transport pyrimidine nucleoside transport sodium ion transmembrane transport pyrimidine-containing compound transmembrane transport nucleoside transmembrane transport purine nucleobase transmembrane transport retina homeostasis purine nucleobase transport transport
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline