Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSYPGYPPTG YPPFPGYPPA GQESSFPPSG QYPYPSGFPP MGGGAYPQVP SSGYPGAGGY PAPGGYPAPG GYPGAPQPGG APSYPGVPPG QGFGVPPGGA GFSGYPQPPS QSYGGGPAQV PLPGGFPGGQ MPSQYPGGQP TYPSQPATVT QVTQGTIRPA ANFDAIRDAE ILRKAMKGFG TDEQAIVDVV ANRSNDQRQK IKAAFKTSYG KDLIKDLKSE LSGNMEELIL ALFMPPTYYD AWSLRKAMQG AGTQERVLIE ILCTRTNQEI REIVRCYQSE FGRDLEKDIR SDTSGHFERL LVSMCQGNRD ENQSINHQMA QEDAQRLYQA GEGRLGTDES CFNMILATRS FPQLRATMEA YSRMANRDLL SSVSREFSGY VESGLKTILQ CALNRPAFFA ERLYYAMKGA GTDDSTLVRI VVTRSEIDLV QIKQMFAQMY QKTLGTMIAG DTSGDYRRLL LAIVGQ
Calcium/phospholipid-binding protein which promotes membrane fusion and is involved in exocytosis. Annexin VII is a member of the annexin family of calcium-dependent phospholipid binding proteins. The Annexin VII gene contains 14 exons and spans approxima
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Annexin A7; Annexin VII; Annexin-7; ANXA7; Interleukin-8; RP11-537A6.8; Synexin; unnamed protein product
Gene Aliases: ANX7; ANXA7; OK/SW-cl.95; SNX; SYNEXIN
UniProt ID: (Human) P20073
Entrez Gene ID: (Human) 310
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support