Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MIHSLFLIN SSGDIFLEKH WKSVVSRSVC DYFFEAQERA TEAENVPPVI PTPHHYLLSV YRHKIFFVAV IQTEVPPLFV IEFLHRVVDT FQDYFGVCSE PVIKDNVVVV YEVLEEMLDN GFPLATESNI LKELIKPPTI LRTVVNTITG STNVGDQLPT GQLSVVPWRR TGVKYTNNEA YFDVIEEIDA IIDKSGSTIT AEIQGVIDAC VKLTGMPDLT LSFMNPRLLD DVSFHPCVRF KRWESERILS FIPPDGNFRL LSYHVSAQKC CLGM (1-273 aa encoded by BC056257)
This gene encodes a subunit of the heterotetrameric adaptor-related protein complex 3 (AP-3), which belongs to the adaptor complexes medium subunits family. The AP-3 complex plays a role in protein trafficking to lysosomes and specialized organelles. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adapter-related protein complex 3 mu-2 subunit; adapter-related protein complex 3 subunit mu-2; adaptor related protein complex 3 mu 2 subunit; adaptor-related protein complex 3 subunit mu-2; adaptor-related protein complex 3, mu 2 subunit; cla; clathrin assembly protein assembly protein complex 1 medium chain homolog 2; clathrin assembly protein assembly protein complex 3 mu-2 medium chain; clathrin coat assembly protein AP47 homolog 2; clathrin coat-associated protein AP47 homolog 2; clathrin-associated protein AP47 homolog 2; epididymis secretory sperm binding protein; golgi adaptor AP-1 47 kDA protein homolog 2; HA1 47 kDa subunit homolog 2; HA1 47kDA subunit homolog 2; m3B; mu3B-adaptin; P47B; unnamed protein product
Gene Aliases: 5830445E16Rik; AP-3B; AP47B; CLA20; P47B
UniProt ID: (Human) P53677, (Rat) P53678, (Mouse) Q8R2R9
Entrez Gene ID: (Human) 10947, (Rat) 140667, (Mouse) 64933
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support