Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ARHGEF7 Antibodies

          Invitrogen

          ARHGEF7 Polyclonal Antibody

          View all (12) ARHGEF7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ARHGEF7 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ARHGEF7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARHGEF7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARHGEF7 Antibody (PA5-88071) in WB

          Western blot analysis of ARHGEF7 in extracts of various cell lines. Samples were incubated with ARHGEF7 Polyclonal antibody (Product # PA5-88071) using a dilution of 1:500, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARHGEF7 Antibody in Western Blot (WB)
          ARHGEF7 Polyclonal Antibody

          Product Details

          PA5-88071

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ARHGEF7 (NP_0011069841)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.48 mg/mL

          Amount

          148 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2804629

          Product Specific Information

          Immunogen sequence: MNSAEQTVTW LITLGVLESP KKTISDPEGF LQASLKDGVV LCRLLERLLP GTIEKTFDAN DLYQGQNFNK VLSSLVTLNK VTADIGLGSD SVCARPSSHR IKSFDSLGSQ SLHTRTSKLF QGQYRSLDMT DNSNNQLVVR AKFNFQQTNE DELSFSKGDV IHVTRVEEGG WWEGTLNGRT GWFPSNYVRE VKASEKPVSP KSGTLKSPPK GFDTTAINKS YYNVVLQNIL ETENEYSKEL QTVLSTYLRP LQTSEKLSSA; Positive Samples: SW620, Jurkat, COS-1, COS-7, MCF7, K-562, Mouse lung, Mouse heart; Cellular Location: Cell junction, Cell projection, Cytoplasm, cell cortex, focal adhesion, lamellipodium, ruffle

          Target Information

          The serine/threonine kinase, p21 activated kinase (PAK), is a downstream effector of the small GTPases Cdc42 and Rac. PAK associates with Nck, the p85 and p110 subunits of PI3-kinase and PIX (PAK-interacting exchange factor) in a focal complex. The binding of PIX is necessary for the localization and activation of PAK in the Cdc42 to Rac signaling pathway, and this binding occurs through the high affinity of the N-terminal SH3 domain of PIX for a conserved proline rich PAK sequence. PIX exists as two isoforms, alpha and beta, and both are highly expressed in heart, muscle and thymus tissues of human and rat. alphaPIX is phosphorylated via PDGF and EphB2 receptor signaling pathways or through association with PI3-kinase. The alphaPIX isoform predominantly acts as a guanine nucleotide exchange factor (GEF) on Rac, which may mediate lamellipodia formation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ARHGEF7; Beta-Pix; COOL-1; KIAA0142; p85; p85SPR; PAK-interacting exchange factor beta; Pak3 binding protein; PIXbeta; Rho guanine nucleotide exchange factor (GEF) 7; Rho guanine nucleotide exchange factor 7; SH3 domain-containing proline-rich protein; unnamed protein product

          View more View less

          Gene Aliases: ARHGEF7; BETA-PIX; betaPix; betaPix-b; betaPix-c; Cool; COOL-1; COOL1; KIAA0142; mKIAA0142; Nbla10314; P50; P50BP; P85; P85COOL1; P85SPR; PAK3; PAK3BP; PIX; PIXB

          View more View less

          UniProt ID: (Human) Q14155, (Mouse) Q9ES28

          View more View less

          Entrez Gene ID: (Human) 8874, (Mouse) 54126

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity protein binding protein kinase binding gamma-tubulin binding Rho guanyl-nucleotide exchange factor activity guanyl-nucleotide exchange factor
          Process(es)
          Golgi organization signal transduction Rho protein signal transduction nervous system development positive regulation of fibroblast migration lamellipodium assembly intracellular signal transduction positive regulation of apoptotic process positive regulation of GTPase activity ephrin receptor signaling pathway focal adhesion assembly positive regulation of substrate adhesion-dependent cell spreading negative regulation of microtubule nucleation positive regulation of lamellipodium morphogenesis hematopoietic progenitor cell differentiation small GTPase mediated signal transduction positive regulation of protein binding regulation of Rho protein signal transduction astrocyte cell migration positive regulation of growth hormone secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0