Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ATP1A2 Antibodies

          Proteintech

          ATP1A2 Polyclonal Antibody, Biotin

          View all (8) ATP1A2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ATP1A2 Polyclonal Antibody, Biotin

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ATP1A2 Antibody (BIOTIN-55179) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse heart tissue slide using Biotin-55179 (ATP1A2-Specific antibody) at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ATP1A2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          BIOTIN-55179

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide

          Conjugate

          Biotin Biotin Biotin

          Form

          Liquid

          Concentration

          1000 µg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Target Information

          Atp1a2 encodes an Na+/K+ ATPase, which is a membrane protein that is composed of two subunits - alpha and beta. The Na+/K+ ATPase pump maintains an essential electrochemical gradient within cells through the active transport of sodium (Na+) and potassium (K+) ions. The Atp1a2 encoded protein belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These ion gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. The ATPase enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta), and the catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. The Atp1a2 gene encodes an alpha 2 subunit. Mutations in the Atp1a2 gene result in familial basilar or hemiplegic migraines, and in a rare syndrome known as alternating hemiplegia of childhood. Diseases associated with ATP1A2 include Alternating Hemiplegia Of Childhood and Migraine, Familial Hemiplegic, 2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ATPase Na+/K+ transporting alpha 2 polypeptide; Na(+)/K(+) ATPase alpha(+) subunit; Na(+)/K(+) ATPase alpha-2 subunit; Na+/K+ ATPase, alpha-A(+) catalytic polypeptide; Na+/K+ ATPase, alpha-B polypeptide; Na,K-ATPase subunit alpha-B; Sodium Potassium ATPase; Sodium pump subunit alpha-2; sodium-potassium ATPase catalytic subunit alpha-2; sodium/potassium-transporting ATPase alpha-2 chain; Sodium/potassium-transporting ATPase subunit alpha-2; unnamed protein product

          View more View less

          Gene Aliases: ATP1A2; Atpa-3; AW060654; DEE98; FARIMPD; FHM2; KIAA0778; MHP2; mKIAA0778

          View more View less

          UniProt ID: (Human) P50993, (Mouse) Q6PIE5

          View more View less

          Entrez Gene ID: (Human) 477, (Mouse) 98660

          View more View less

          Function(s)
          nucleotide binding sodium:potassium-exchanging ATPase activity steroid binding protein binding ATP binding potassium-transporting ATPase activity phosphatase activity ATPase activity cation-transporting ATPase activity potassium ion binding sodium ion binding metal ion binding protein heterodimerization activity chaperone binding steroid hormone binding hydrolase activity primary active transporter
          Process(es)
          neurotransmitter uptake behavioral fear response regulation of the force of heart contraction regulation of respiratory gaseous exchange by neurological system process ion transport potassium ion transport sodium ion transport cellular sodium ion homeostasis regulation of muscle contraction regulation of smooth muscle contraction regulation of striated muscle contraction cell communication blood circulation regulation of heart contraction regulation of blood pressure adult locomotory behavior visual learning establishment or maintenance of transmembrane electrochemical gradient regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion response to auditory stimulus neuronal action potential propagation regulation of vasoconstriction L-ascorbic acid metabolic process amygdala development olfactory cortex development cellular cation homeostasis cellular potassium ion homeostasis response to nicotine locomotory exploration behavior sodium ion transmembrane transport response to potassium ion sodium ion export from cell locomotion fear response negative regulation of heart contraction positive regulation of heart contraction negative regulation of striated muscle contraction ATP metabolic process negative regulation of cytosolic calcium ion concentration membrane depolarization regulation of glutamate uptake involved in transmission of nerve impulse regulation of synaptic transmission, glutamatergic relaxation of cardiac muscle cardiac muscle contraction cellular response to mechanical stimulus cellular response to steroid hormone stimulus potassium ion transmembrane transport regulation of cardiac muscle cell contraction membrane repolarization membrane depolarization during cardiac muscle cell action potential cell communication by electrical coupling involved in cardiac conduction cation transmembrane transport transport across blood-brain barrier hydrogen ion transmembrane transport negative regulation of calcium ion transmembrane transport response to glycoside potassium ion import across plasma membrane transport potassium ion import ATP hydrolysis coupled proton transport negative regulation of calcium:sodium antiporter activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline