Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • AXIN1 Antibodies

          Rockland

          AXIN1 Polyclonal Antibody

          View all (28) AXIN1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AXIN1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          AXIN1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AXIN1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AXIN1 Antibody (600-401-Z14) in ICC/IF

          Immunofluorescence of AXIN1. Tissue: human brain tissue. Primary Antibody: Anti-AXIN1 antibody at 20 μg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AXIN1 Antibody in Immunocytochemistry (ICC/IF)
          AXIN1 Antibody in Western Blot (WB)
          AXIN1 Polyclonal Antibody

          Product Details

          600-401-Z14

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          20 µg/mL
          -

          ELISA (ELISA)

          1:10,000-1:20,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Anti-AXIN1 antibody was prepared from whole rabbit serum produced by repeated immunizations with a 14 amino acid synthetic peptide near the C-terminus of human AXIN1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          AXIN1 is a cytoplasmic protein which contains a regulation of G-protein signaling (RGS) domain and a disheveled and axin (DIX) domain and is thought to function as a negative regulator of the WNT signaling pathway that regulates embryonic axis formation. AXIN1 interacts with adenomatosis polyposis coli (APC), beta-catenin, glycogen synthase kinase 3 beta, forming a tetrameric complex resulting in the regulation of the stabilization of beta-catenin. Mutations in the AXIN1 gene have been associated various carcinomas, indicating that it also functions as a tumor suppressor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Axin-1; Axis inhibition protein 1; axis inhibitor 1; contains RGS domain and Daughterless similarity; fused, mouse, homolog of; GSK-3beta interacting protein rAxin; hAxin; LA16c-314G4.3; MGC52315; Protein Fused; protein phosphatase 1, regulatory subunit 49; rAxin

          View more View less

          Gene Aliases: AI316800; AXIN; AXIN1; CMDOH; Fu; fused; Kb; Ki; kinky; knobbly; PPP1R49

          View more View less

          UniProt ID: (Human) O15169, (Rat) O70239, (Mouse) O35625

          View more View less

          Entrez Gene ID: (Human) 8312, (Rat) 79257, (Mouse) 12005

          View more View less

          Function(s)
          p53 binding protein binding beta-catenin binding enzyme binding protein kinase binding protein domain specific binding ubiquitin protein ligase binding signaling adaptor activity identical protein binding protein homodimerization activity protein serine/threonine kinase activator activity SMAD binding binding, bridging armadillo repeat domain binding I-SMAD binding R-SMAD binding protein serine/threonine kinase binding protein binding, bridging involved in substrate recognition for ubiquitination signal transducer activity GTPase activator activity receptor binding protein C-terminus binding receptor signaling complex scaffold activity protein complex scaffold protein self-association scaffold/adaptor protein
          Process(es)
          protein polyubiquitination in utero embryonic development nucleocytoplasmic transport apoptotic process sensory perception of sound dorsal/ventral axis specification dorsal/ventral pattern formation negative regulation of gene expression Wnt signaling pathway protein catabolic process negative regulation of Wnt signaling pathway positive regulation of transforming growth factor beta receptor signaling pathway cytoplasmic microtubule organization positive regulation of protein ubiquitination positive regulation of proteasomal ubiquitin-dependent protein catabolic process negative regulation of transcription elongation from RNA polymerase II promoter post-anal tail morphogenesis proteasome-mediated ubiquitin-dependent protein catabolic process epigenetic programming in the zygotic pronuclei negative regulation of fat cell differentiation positive regulation of protein catabolic process positive regulation of JNK cascade axial mesoderm development axial mesoderm formation cell development negative regulation of protein metabolic process canonical Wnt signaling pathway head development regulation of canonical Wnt signaling pathway protein-containing complex assembly negative regulation of canonical Wnt signaling pathway beta-catenin destruction complex assembly positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process optic placode formation regulation of protein phosphorylation positive regulation of protein phosphorylation activation of JUN kinase activity determination of left/right symmetry adult walking behavior cell death hemoglobin metabolic process forebrain anterior/posterior pattern specification Wnt-activated signaling pathway involved in forebrain neuron fate commitment olfactory placode formation activation of protein kinase activity erythrocyte homeostasis regulation of catenin import into nucleus termination of G-protein coupled receptor signaling pathway positive regulation of JUN kinase activity positive regulation of GTPase activity cellular protein complex assembly positive regulation of protein kinase activity positive regulation of transcription, DNA-templated embryonic eye morphogenesis neurological system process protein homooligomerization positive regulation of ubiquitin-protein transferase activity muscle cell development embryonic skeletal joint morphogenesis canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation cellular response to organic cyclic compound genetic imprinting Wnt signaling pathway involved in somitogenesis signal transduction multicellular organism development cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline