Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAMVSEFLK QAWFIENEEQ EYVQTVKSSK GGPGSAVSPY PTFNPSSDVA ALHKAIMVKG VDEATIIDIL TKRNNAQRQQ IKAAYLQETG KPLDETLKKA LTGHLEEVVL ALLKTPAQFD ADELRAAMKG LGTDEDTLIE ILASRTNKEI RDINRVYREE LKRDLAKDIT SDTSGDFRNA LLSLAKGDRS EDFGVNEDLA DSDARALYEA GERRKGTDVN VFNTILTTRS YPQLRRVFQK YTKYSKHDMN KVLDLELKGD IEKCLTAIVK CATSKPAFFA EKLHQAMKGV GTRHKALIRI MVSRSEIDMN DIKAFYQKMY GISLCQAILD ETKGDYEKIL VALCGGN (1-346 aa encoded by BC001275)
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin I; annexin I (lipocortin I); annexin-1; ANX1; ANXA1; ANXA1; calcium and phospholipid-binding protein; calpactin II; calpactin-2; chromobindin-9; epididymis secretory sperm binding protein; lipocortin 1; lipocortin I; lipocortin I protein; LPC1; p35; phospholipase A2 inhibitory protein; RP11-71A24.1; unnamed protein product
Gene Aliases: Anx-1; Anx-A1; ANX1; C430014K04Rik; Lpc-1; LPC1; p35
UniProt ID: (Human) P04083, (Rat) P07150, (Mouse) P10107
Entrez Gene ID: (Human) 301, (Rat) 25380, (Mouse) 16952
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support