Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
BAGE2 (BAGE Family Member 2) is a Protein Coding gene. Candidate gene encoding tumor antigens.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: member of BAGE family; tumor antigen; unnamed protein product
Gene Aliases: CT2.2
Entrez Gene ID: (Human) 85319
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support