Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BBS12 Antibodies

          Abnova

          BBS12 Polyclonal Antibody, MaxPab™

          View all (4) BBS12 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BBS12 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          BBS12 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BBS12 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BBS12 Antibody (H00166379-B01P) in WB

          FLJ35630 MaxPab polyclonal antibody. Western Blot analysis of FLJ35630 expression in Y-79. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BBS12 Antibody in Western Blot (WB)
          BBS12 Polyclonal Antibody, MaxPab™

          Product Details

          H00166379-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BBS12 (NP_689831.2, 1 a.a. approximately 710 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVMACRVVNK RRHMGLQQLS SFAETGRTFL GPLKSSKFII DEECHESVLI SSTVRLLESL DLTSAVGQLL NEAVQAQNNT YRTGISTLLF LVGAWSSAVE ECLHLGVPIS IIVSVMSEGL NFCSEEVVSL HVPVHNIFDC MDSTKTFSQL ETFSVSLCPF LQVPSDTDLI EELHGLKDVA SQTLTISNLS GRPLKSYELF KPQTKVEADN NTSRTLKNSL LADTCCRQSI LIHSRHFNRT DNTEGVSKPD GFQEHVTATH KTYRCNDLVE LAVGLSHGDH SSMKLVEEAV QLQYQNACVQ QGNCTKPFMF DISRIFTCCL PGLPETSSCV CPGYITVVSV SNNPVIKELQ NQPVRIVLIE GDLTENYRHL GFNKSANIKT VLDSMRLQED SSEELWANHV LQVLIQFKVN LVLVQGNVSE RLIEKCINSK RLVIGSVNGS VMQAFAEAAG AVQVAYITQV NEDCVGDGVC VTFWRSSPLD VVDRNNRIAI LLKTEGINLV TAVLTNPVTA QMQIKEDRFW TCAYRLYYAL KEEKVFLGGG AVEFLCLSCL HILAEQSLKK ENHACSGWLH NTSSWLASSL AIYRPTVLKF LANGWQKYLS TLLYNTANYS SEFEASTYIQ HHLQNATDSG SPSSYILNEY SKLNSRIFNS DISNKLEQIP RVYDVVTPKI EAWRRALDLV LLVLQTDSEI ITGHGHTQIN SQELTGFLFL

          Target Information

          The BBS12 gene is a key contributor to Bardet-Biedl Syndrome (BBS), a rare genetic disorder characterized by primary cilia dysfunction, leading to diverse clinical manifestations such as retinal degeneration, obesity, polydactyly, renal abnormalities, and learning difficulties. BBS12 is located on chromosome 4q27 and encodes a vertebrate-specific chaperonin-like protein. Together with BBS6 and BBS10, BBS12 forms a part of a complex associated with the CCT/TRiC family of chaperonins, which are essential for the proper assembly of the BBSome complex. This complex plays a critical role in the trafficking of vesicles to cilia and is crucial for ciliary function and structure. Mutations in BBS12 are responsible for approximately 5% of BBS cases, underscoring its significant role in the pathophysiology of the syndrome. These mutations often lead to disruptions in ciliary functions, which are pivotal for the syndrome's manifestation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Bardet-Biedl syndrome 12 protein; Bardet-Biedl syndrome 12 protein homolog; Chaperonin-containing T-complex member BBS12; DN-452A22.3; unnamed protein product

          View more View less

          Gene Aliases: BBS12; C4orf24; Gm1805; Gm407; Gm721

          View more View less

          UniProt ID: (Human) Q6ZW61, (Mouse) Q5SUD9

          View more View less

          Entrez Gene ID: (Human) 166379, (Mouse) 241950

          View more View less

          Function(s)
          protein binding ATP binding molecular_function
          Process(es)
          intraciliary transport eating behavior photoreceptor cell maintenance negative regulation of fat cell differentiation chaperone-mediated protein complex assembly fat cell differentiation stem cell differentiation negative regulation of stem cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline