Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MSERRVVVD LPTSASSSMP LQRRRASFRG PRSSSSLESP PASRTNAMSG LVRAPGVYVG TAPSGCIGGL GARVTRRALG ISSVFLQGLR SSGLATVPAP GLERDHGAVE DLGGCLVEYM AKVHALEQVS QELETQLRMH LESKATRSGN WGALRASWAS SCQQVGEAVL ENARLMLQTE TIQAGADDFK ERYENEQPFR KAAEEEINSL YKVIDEANLT KMDLES (1-225 aa encoded by B C113518)
More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, the protein product of this gene (phakinin), and filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene have been associated with juvenile-onset, progressive cataracts and Dowling-Meara epidermolysis bullosa simplex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 49 kDa cytoskeletal protein; beaded filament protein CP49; beaded filament structural protein 2, phakinin; bfps2, Cytoskeletal protein, 49-kD; lens fiber cell beaded filament protein CP 47; lens fiber cell beaded filament protein CP 49; lens intermediate filament-like light; MGC142078; MGC142080; unnamed protein product
Gene Aliases: CP47; CP49; CTRCT12; LIFL-L; PHAKOSIN
UniProt ID: (Human) Q13515, (Mouse) Q6NVD9, (Rat) D3ZER2
Entrez Gene ID: (Human) 8419, (Mouse) 107993, (Rat) 501046
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support