Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This antibody shows 100% cross-reactivity with recombinant zebra fish (rzf) BMP-2 and rhBMP-4, 20%-40% cross-reactivity with rhBMP-3, -3b, -6, and no cross-reactivity with rhBMP-5, -10, or -15.
Reconstitute at 0.5 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
Western blot: 1 µg/mL, under non--reducing conditions only
Neutralization: Measured by its ability to neutralize BMP¬2-induced alkaline phosphatase production in the ATDC5 mouse chondrogenic cell line. The Neutralization Dose (ND50) is typically 5-15 µg/mL in the presence of 1 µg/mL Recombinant Human BMP-2 and 2 µg/mL heparin. This antibody also neutralizes Human BMP-4 induced activity in the ATDC5 mouse chondrogenic cell line. The ND50 is approximately 3 µg/mL in the presence of 75 ng/mL of Recombinant Human BMP-4.
Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP-2; BMP-2A; BMP-2B; BMP-4; Bone morphogenetic protein 2; Bone morphogenetic protein 2A; Bone morphogenetic protein 2B; Bone morphogenetic protein 4; osteogenic protein 2; unnamed protein product
Gene Aliases: BDA2; BMP2; BMP2A; BMP2B; BMP2B1; BMP4; DVR4; MCOPS6; OFC11; SSFSC; SSFSC1; ZYME
UniProt ID: (Human) P12643, (Human) P12644
Entrez Gene ID: (Human) 650, (Human) 652
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support