Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BMP-7 Antibodies

          Invitrogen

          BMP-7 Polyclonal Antibody, Biotin, PeproTech®

          View all (25) BMP-7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BMP-7 Polyclonal Antibody, Biotin, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          BMP-7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BMP-7 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMP-7 Antibody (500-P198BT-50UG) in WB

          Western Blot: To detect Human BMP-7 by Western Blot analysis BMP-7 Polyclonal Antibody, Biotin (Product # 500-P198BT-500UG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BMP-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMP-7 Antibody in Western Blot (WB)
          BMP-7 Antibody in Western Blot (WB)
          BMP-7 Antibody in ELISA (ELISA)
          BMP-7 Polyclonal Antibody, Biotin, PeproTech®

          Product Details

          500-P198BT-50UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          -

          ELISA (ELISA)

          0.25-1.0 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CHO cells-derived Recombinant Human BMP-7
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          50 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2929779

          Product Specific Information

          AA Sequence of recombinant protein: MANVAENSSS DQRQACKKHE LYVSFRDLGW QDWIIAPEGY AAYYCEGECA FPLNSYMNAT NHAIVQTLVH FINPETVPKP CCAPTQLNAI SVLYFDDSSN VILKKYRNMV VRACGCH.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BMP-7. Anti-Human BMP-7-specific antibody was purified by affinity chromatography and then biotinylated.

          Sandwich ELISA: To detect hBMP-7 by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BMP-7 (500-P198) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBMP-7.

          Western Blot: To detect Human BMP-7 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human BMP-7 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          500-P198BT-1MG will be provided as 2 x 500 µg

          Target Information

          The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BMP; BMP-7; BMP7; Bone morphogenetic protein; Bone morphogenetic protein 7; OP-1; Osteogenic protein 1; RP11-560A15.5; unnamed protein product

          View more View less

          Gene Aliases: BMP7; OP-1; OP1

          View more View less

          UniProt ID: (Human) P18075

          View more View less

          Entrez Gene ID: (Human) 655

          View more View less

          Function(s)
          cytokine activity protein binding growth factor activity heparin binding protein serine/threonine kinase activator activity receptor agonist activity BMP receptor binding
          Process(es)
          skeletal system development ossification osteoblast differentiation eye development metanephros development ureteric bud development mesoderm formation kidney development mesonephros development epithelial to mesenchymal transition endocardial cushion formation pericardium morphogenesis pattern specification process axon guidance salivary gland morphogenesis heart development cell proliferation negative regulation of cell proliferation embryonic pattern specification organ morphogenesis positive regulation of gene expression negative regulation of striated muscle cell apoptotic process positive regulation of epithelial to mesenchymal transition dendrite development neural fold elevation formation cell differentiation embryonic limb morphogenesis positive regulation of bone mineralization BMP signaling pathway epithelial cell differentiation positive regulation of epithelial cell differentiation hindbrain development response to estradiol response to vitamin D positive regulation of heterotypic cell-cell adhesion tube morphogenesis ameloblast differentiation odontogenesis of dentin-containing tooth positive regulation of apoptotic process response to peptide hormone positive regulation of cell differentiation negative regulation of neuron differentiation positive regulation of neuron differentiation positive regulation of osteoblast differentiation negative regulation of Notch signaling pathway negative regulation of cell cycle negative regulation of mitotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cell development camera-type eye morphogenesis embryonic camera-type eye morphogenesis anatomical structure formation involved in morphogenesis cardiac muscle tissue development branching morphogenesis of an epithelial tube mesenchymal cell differentiation neuron projection morphogenesis negative regulation of neurogenesis cartilage development positive regulation of multicellular organismal process pharyngeal system development embryonic skeletal joint morphogenesis positive regulation of SMAD protein import into nucleus cardiac septum morphogenesis branching involved in salivary gland morphogenesis mesenchyme development negative regulation of prostatic bud formation regulation of branching involved in prostate gland morphogenesis chorio-allantoic fusion heart trabecula morphogenesis monocyte aggregation cellular response to hypoxia cellular response to BMP stimulus negative regulation of mesenchymal cell apoptotic process involved in nephron morphogenesis negative regulation of glomerular mesangial cell proliferation metanephric mesenchyme morphogenesis nephrogenic mesenchyme morphogenesis metanephric mesenchymal cell proliferation involved in metanephros development positive regulation of brown fat cell differentiation cell-cell adhesion positive regulation of dendrite development positive regulation of hyaluranon cable assembly mesenchymal cell apoptotic process involved in nephron morphogenesis negative regulation of NIK/NF-kappaB signaling allantois development positive regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis regulation of multicellular organismal development regulation of removal of superoxide radicals
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline