Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BMP-2 Antibodies

          Abnova

          BMP2 Recombinant Rabbit Monoclonal Antibody

          View all (45) BMP-2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BMP2 Recombinant Rabbit Monoclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          BMP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BMP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMP2 Antibody (RAB01554) in WB

          Western blot analysis of BMP2 in MCF-7, Hela lysates using human BMP2 recombinant monoclonal antibody, clone R03-3H9 (Product # RAB01554). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMP2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          RAB01554

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Expression System

          CHO cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to human BMP2.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          50mM tris glycine, pH 7.4, with 0.05% BSA, 40% glycerol, 150mM NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Bone Morphogenic Proteins (BMP) are members of the TGF-beta superfamily that affect bone and cartilage formation (Hogan 1996, Reddi 1998 and Francis-West et al. 1999). Mature BMPs are 30-38 kDa proteins that assume a TGF-beta -like cysteine knot configuration. Lovostatin increases bone formation by turning on the bmp-2 gene (Mundy et al. 1999). BMPs stimulate the production of specific bone matrix proteins and alter stromal cell and osteoclast proliferation (Macias et al. 1999, Lecanda et al. 1997). BMPs may also be an important factor for development of the viscera, with roles in cell proliferation, apoptosis, differentiation, and morphogenesis (Hogan 1996, Dale and Wardle 1999). BMPs appear to be responsible for normal dorsal/ventral patterning. Like TGF-beta, BMPs bind to a type II receptor, which then recruits the transducing type I receptor unit, activating the Smad protein signaling pathway (Massague 1994, Derynck 1997, Attisano 1993).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2610024H22Rik; AL117858; AW546137; BB189135; BMP; BMPR-II; BMPRII; Bone morphogenetic; Bone morphogenetic protein; bone morphogenetic protein 2A; H-BMP-2; osteogenic protein 2

          View more View less

          Gene Aliases: BDA2; BMP2A; SSFSC; SSFSC1

          View more View less

          UniProt ID: (Human) P12643

          View more View less

          Entrez Gene ID: (Human) 650

          View more View less

          Function(s)
          receptor binding cytokine activity protein binding growth factor activity phosphatase activator activity co-receptor binding receptor agonist activity BMP receptor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development osteoblast differentiation branching involved in ureteric bud morphogenesis response to hypoxia in utero embryonic development epithelial to mesenchymal transition positive regulation of protein phosphorylation chondrocyte differentiation myeloid leukocyte differentiation heart morphogenesis heart induction endodermal-mesodermal cell signaling aortic valve development atrioventricular valve morphogenesis endocardial cushion morphogenesis cardiac atrium formation endocardial cushion formation positive regulation of extracellular matrix constituent secretion regulation of transcription, DNA-templated cellular iron ion homeostasis inflammatory response Notch signaling pathway cell-cell signaling heart development positive regulation of cell proliferation negative regulation of cell proliferation organ morphogenesis gene expression positive regulation of gene expression negative regulation of gene expression positive regulation of epithelial to mesenchymal transition negative regulation of steroid biosynthetic process positive regulation of phosphatase activity telencephalon development telencephalon regionalization negative regulation of cell-cell adhesion positive regulation of Wnt signaling pathway bone mineralization osteoclast differentiation positive regulation of cell migration positive regulation of bone mineralization BMP signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of aldosterone biosynthetic process positive regulation of osteoblast proliferation cardiocyte differentiation embryonic heart tube anterior/posterior pattern specification positive regulation of peroxisome proliferator activated receptor signaling pathway odontogenesis of dentin-containing tooth positive regulation of odontogenesis positive regulation of apoptotic process positive regulation of MAPK cascade negative regulation of insulin-like growth factor receptor signaling pathway cell fate commitment positive regulation of cell differentiation negative regulation of fat cell differentiation positive regulation of fat cell differentiation positive regulation of neuron differentiation positive regulation of ossification negative regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of smooth muscle cell proliferation positive regulation of astrocyte differentiation mesenchymal cell differentiation inner ear development negative regulation of calcium-independent cell-cell adhesion cardiac muscle cell differentiation cardiac muscle tissue morphogenesis pericardium development corticotropin hormone secreting cell differentiation thyroid-stimulating hormone-secreting cell differentiation cardiac epithelial to mesenchymal transition bone development positive regulation of SMAD protein import into nucleus lung vasculature development mesenchyme development muscle tissue development positive regulation of cartilage development positive regulation of ERK1 and ERK2 cascade cellular response to BMP stimulus mesenchymal cell proliferation involved in ureteric bud development negative regulation of canonical Wnt signaling pathway positive regulation of bone mineralization involved in bone maturation positive regulation of p38MAPK cascade positive regulation of odontoblast differentiation positive regulation of pri-miRNA transcription from RNA polymerase II promoter cardiac jelly development atrioventricular canal morphogenesis negative regulation of cortisol biosynthetic process regulation of cardiac muscle cell differentiation negative regulation of cardiac muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch