    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • V5 Tag Antibodies
          Bioss
          V5 Tag Monoclonal Antibody (5D7), HRP
          View all (83) V5 Tag antibodies
          Questions & Answers
          Cite V5 Tag Monoclonal Antibody (5D7), HRP
          Additional Information:
          {{banner.description}}
          Product Details
          BSM-33005M-HRP
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:300-1:5,000
          -
          ELISA (ELISA)
          1:500-1,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5D7
          Immunogen
          KLH conjugated V5-tag peptide
          Conjugate
          HRP HRP HRP
          • Unconjugated
          • Biotin 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4, with 1% BSA, 50% glycerol
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C or -80°C if preferred
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
          Bioinformatics
          Protein Aliases: simian virus 5 antibody; SPV5gp2; sv5; v-5; V5; V5 Epitope Tag
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline