    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SARS-CoV-2 Nucleocapsid Antibodies
          Bioss
          SARS-CoV-2 Nucleocapsid Protein Monoclonal Antibody (1C7), HRP
          View all (182) SARS-CoV-2 Nucleocapsid antibodies
          Questions & Answers
          Cite SARS-CoV-2 Nucleocapsid Protein Monoclonal Antibody (1C7), HRP
          Additional Information:
          {{banner.description}}
          Product Details
          BSM-41411M-HRP
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:300-1:5,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:400
          -
          ELISA (ELISA)
          1:5,000-10,000
          -
          Product Specifications
          Species Reactivity
          Virus
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1C7
          Immunogen
          Recombinant SARS-CoV-2 N protein (His-tag)
          Conjugate
          HRP HRP HRP
          • Unconjugated
          • Biotin 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          See Datasheet
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          TBS, pH 7.4, with 50% glycerol, 1% BSA
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C or -80°C if preferred
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
          Bioinformatics
          Protein Aliases: COVID-19 nucleocapsid; COVID19 Nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline