Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Bub1 Antibodies

          Abnova

          BUB1 Polyclonal Antibody, MaxPab™

          View all (24) Bub1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite BUB1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          BUB1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BUB1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BUB1 Antibody (H00000699-D01P) in ICC/IF

          Immunofluorescence of purified MaxPab antibody to BUB1 on HeLa cell. Antibody concentration is 10 µg/mL. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BUB1 Antibody in Immunocytochemistry (ICC/IF)
          BUB1 Antibody in Western Blot (WB)
          BUB1 Antibody in Western Blot (WB)
          BUB1 Antibody in Western Blot (WB)
          BUB1 Antibody in Proximity Ligation Assay (PLA) (PLA)
          BUB1 Polyclonal Antibody, MaxPab™

          Product Details

          H00000699-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10 µg/mL
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BUB1 (NP_004327.1, 1 a.a. approximately 1085 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MDTPENVLQM LEAHMQSYKG NDPLGEWERY IQWVEENFPE NKEYLITLLE HLMKEFLDKK KYHNDPRFIS YCLKFAEYNS DLHQFFEFLY NHGIGTLSSP LYIAWAGHLE AQGELQHASA VLQRGIQNQA EPREFLQQQY RLFQTRLTET HLPAQARTSE PLHNVQVLNQ MITSKSNPGN NMACISKNQG SELSGVISSA CDKESNMERR VITISKSEYS VHSSLASKVD VEQVVMYCKE KLIRGESEFS FEELRAQKYN QRRKHEQWVN EDRHYMKRKE ANAFEEQLLK QKMDELHKKL HQVVETSHED LPASQERSEV NPARMGPSVG SQQELRAPCL PVTYQQTPVN MEKNPREAPP VVPPLANAIS AALVSPATSQ SIAPPVPLKA QTVTDSMFAV ASKDAGCVNK STHEFKPQSG AEIKEGCETH KVANTSSFHT TPNTSLGMVQ ATPSKVQPSP TVHTKEALGF IMNMFQAPTL PDISDDKDEW QSLDQNEDAF EAQFQKNVRS SGAWGVNKII SSLSSAFHVF EDGNKENYGL PQPKNKPTGA RTFGERSVSR LPSKPKEEVP HAEEFLDDST VWGIRCNKTL APSPKSPGDF TSAAQLASTP FHKLPVESVH ILEDKENVVA KQCTQATLDS CEENMVVPSR DGKFSPIQEK SPKQALSSHM YSASLLRLSQ PAAGGVLTCE AELGVEACRL TDTDAAIAED PPDAIAGLQA EWMQMSSLGT VDAPNFIVGN PWDDKLIFKL LSGLSKPVSS YPNTFEWQCK LPAIKPKTEF QLGSKLVYVH HLLGEGAFAQ VYEATQGDLN DAKNKQKFVL KVQKPANPWE FYIGTQLMER LKPSMQHMFM KFYSAHLFQN GSVLVGELYS YGTLLNAINL YKNTPEKVMP QGLVISFAMR MLYMIEQVHD CEIIHGDIKP DNFILGNGFL EQDDEDDLSA GLALIDLGQS IDMKLFPKGT IFTAKCETSG FQCVEMLSNK PWNYQIDYFG VAATVYCMLF GTYMKVKNEG GECKPEGLFR RLPHLDMWNE FFHVMLNIPD CHHLPSLDLL RQKLKKVFQQ HYTNKIRALR NRLIVLLLEC KRSRK

          Target Information

          The BUB1 gene encodes a serine/threonine-protein kinase that is integral to the spindle assembly checkpoint, a crucial mechanism ensuring proper chromosome alignment and segregation during mitosis. BUB1 is involved in detecting and correcting errors in kinetochore-microtubule attachments, which is vital for maintaining genomic stability. It acts by delaying the onset of anaphase until all chromosomes are correctly attached to the spindle apparatus. BUB1 is recruited to kinetochores in early mitosis, where it phosphorylates several substrates to activate other checkpoint proteins, such as BUBR1 and MAD2, thereby preventing premature progression through mitosis. Mutations or aberrations in the expression levels of BUB1 are often associated with aneuploidy and chromosomal instability, contributing to cancer progression. BUB1 is thus considered a potential target for cancer therapeutics, aiming to exploit its role in the mitotic checkpoint to selectively eliminate tumor cells through checkpoint bypass or mitotic arrest.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BUB1 budding uninhibited by benzimidazoles 1 homolog; BUB1A; budding uninhibited by benzimidazoles 1 homolog; hBUB1; huBUB1; HuBub1; putative serine/threonine-protein kinase; mBUB1; Mitotic checkpoint serine/threonine-protein kinase BUB1; mitotic spindle checkpoint kinase; protein kinase; putative serine/threonine-protein kinase; unnamed protein product

          View more View less

          Gene Aliases: AL022991; BUB1; BUB1A; BUB1L; C80208; D2Xrf87; hBUB1; MCPH30

          View more View less

          UniProt ID: (Human) O43683, (Mouse) O08901

          View more View less

          Entrez Gene ID: (Human) 699, (Mouse) 12235

          View more View less

          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity protein serine kinase activity histone H2A kinase activity transferase activity, transferring phosphorus-containing groups non-receptor serine/threonine protein kinase
          Process(es)
          chromatin remodeling apoptotic process chromosome segregation regulation of sister chromatid cohesion mitotic spindle assembly checkpoint intracellular signal transduction cell division meiotic sister chromatid cohesion, centromeric regulation of chromosome segregation positive regulation of maintenance of mitotic sister chromatid cohesion, centromeric protein phosphorylation cell cycle mitotic nuclear division embryo development phosphorylation spindle assembly checkpoint mitotic spindle checkpoint positive regulation of intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0