Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD157 (BST1) Antibodies

          Fabgennix

          Bone marrow stromal cell antigen 1 Polyclonal Antibody

          View all (36) CD157 (BST1) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Bone marrow stromal cell antigen 1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Bone marrow stromal cell antigen 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Bone marrow stromal cell antigen 1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Bone marrow stromal cell antigen 1 Antibody (BST1-101AP) in WB

          WB of BST1-101AP with BB cells on 10% SDS gel. 1:250 antibody dilution in DiluObuffer for 1 hour at room temperature. Apparent MW is 39 kDa. W, B of BST1-101AP with 1) Chondrocytes and 2) SKNMC cells. 1:500 antibody dilution in DiluObuffer. Apparent MW is 37 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Bone marrow stromal cell antigen 1 Antibody in Western Blot (WB)
          Bone marrow stromal cell antigen 1 Polyclonal Antibody

          Product Details

          BST1-101AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (IHC)

          1:50-1:250
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:250
          -

          ELISA (ELISA)

          1:10,000
          -

          Immunoprecipitation (IP)

          1:50-1:250
          -

          Immunomicroscopy (IM)

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to amino acids 255-274 of Human Bone marrow stromal cell antigen 1. Peptide was covalently modified.Sequence:FQY SCI NDY RPV KLL QCV DH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.5 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          CD157, also known as Bone Marrow Stromal Cell Antigen 1 (BST1), is a pleiotropic ectoenzyme and glycosylphosphatidylinositol-anchored molecule that functions both as an enzyme and a receptor. Similar to CD38, CD157 is involved in the metabolism of NAD+, which may play a role in regulating intracellular Ca2+ levels. As an enzyme, CD157 is also referred to as ADP-ribosyl cyclase 2 and DAMP-1, and it facilitates pre-B cell growth. As a receptor, CD157 is thought to initiate a signal transduction cascade upon binding to its putative ligand, leading to the phosphorylation of cytoplasmic proteins, including focal adhesion kinase (FAK). While the mechanism and functional significance of CD157-initiated signal transduction are not fully characterized, its role in cellular signaling is evident. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis, suggesting a link to polyclonal B cell abnormalities observed in the disease. This overexpression in the stromal cell population may contribute to the pathophysiology of rheumatoid arthritis, highlighting the importance of CD157/BST1 in both normal immune function and disease states.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; Antigen BP3; Bone marrow stromal antigen 1; Bone marrow stromal cell antigen 1; BP-3 alloantigen; BST-1; cADPR hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; Leukocyte antigen 65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase; unnamed protein product

          View more View less

          Gene Aliases: 114/A10; A530073F09; BP-3; Bp3; BST1; Bsta1; cADPR2; CD157; Ly65

          View more View less

          UniProt ID: (Human) Q10588, (Rat) Q63072, (Mouse) Q64277

          View more View less

          Entrez Gene ID: (Human) 683, (Rat) 81506, (Mouse) 12182

          View more View less

          Function(s)
          transferase activity hydrolase activity hydrolase activity, acting on glycosyl bonds phosphorus-oxygen lyase activity NAD+ nucleotidase, cyclic ADP-ribose generating molecular_function NAD+ nucleosidase activity NAD(P)+ nucleosidase activity cyclase lyase metabolite interconversion enzyme
          Process(es)
          regulation of cell-matrix adhesion regulation of cellular extravasation humoral immune response signal transduction positive regulation of cell proliferation positive regulation of B cell proliferation regulation of actin cytoskeleton organization regulation of inflammatory response regulation of calcium-mediated signaling regulation of neutrophil chemotaxis regulation of superoxide metabolic process regulation of integrin-mediated signaling pathway biological_process metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline