Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17476 targets C3a Receptor in FACS and IHC (F) applications and shows reactivity with Rat samples.
The MA5-17476 immunogen is rBL-2H3 transfectants expressing rat C3aR.
C3AR1 is a receptor for the chemotactic and inflammatory peptide anaphylatoxin C3a. The receptor stimulates chemotaxis, granule enzyme release and superoxide anion production. (Uniprot)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3a anaphylatoxin chemotactic receptor; C3a receptor; C3a-R; C3AR
Gene Aliases: C3ar1
UniProt ID: (Rat) O55197
Entrez Gene ID: (Rat) 84007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support