Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEGLEENGGV VQVGELLPCK ICGRTFFPVA LKKHGPICQK TATKKRKTFD SSRQRAEGTD IPTVKPLKPR PEPPKKPSNW RRKHEEFIAT IRAAKGLDQA LKEGGKLPPP PPPSYDPDYI QCPYCQRRFN ENAADRHINF CKEQAARISN KGKFSTDTKG KPTSRTQVYK PPALKKSNSP GTASSGSSRL PQPSGAGKTV VGVPSGKVSS SSSSLGNKLQ TLSPSHKGIA APHAGANVKP RNSTPPSLAR NPAPGVLTNK RKTYTESYIA RPDGDCASSL NGGNIKGIEG HSPGNLPKFC HECGTKYPVE WAKFCCECGI RRMIL
The CGI-62 gene encodes a hypothetical protein located on chromosome 8.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 164, member A; protein FAM164A; unnamed protein product; Zinc finger C2HC domain-containing protein 1A
Gene Aliases: C8orf70; CGI-62; FAM164A; ZC2HC1A
UniProt ID: (Human) Q96GY0
Entrez Gene ID: (Human) 51101
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support