Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: SEEKGAALM RNRDLQLEID QLKHSLMKAE DDCKVERKHT LKLRHAMEQR PSQELLWELQ QEKALLQARV QELEASVQEG KLDRSSPYIQ VLEEDWRQAL RDHQEQANTI FSLRKDLRQG EARRLRCMEE KEMFELQCLA LRKDSKMYKD RIEAILLQME EVAIERDQAI ATREELHAQH ARGLQEKDAL RKQVRELGEK ADELQLQVFQ CEAQLLAVEG RLRRQQLETL VLSSDLEDGS PRRSQELSLP QDLEDTQLSD KGCLAGGGSP KQPFAALHQE QVLRNPHDAG PAGLPGIGAV C (193-492 aa encoded by BC008877)
The protein encoded by this gene is a member of the CARD protein family, which is defined by the presence of a characteristic caspase-associated recruitment domain (CARD). CARD is a protein interaction domain known to participate in activation or suppression of CARD containing members of the caspase family, and thus plays an important regulatory role in cell apoptosis. This protein was identified by its selective association with the CARD domain of BCL10, a postive regulator of apoptosis and NF-kappaB activation, and is thought to function as a molecular scaffold for the assembly of a BCL10 signaling complex that activates NF-kappaB. Several alternatively spliced transcript variants have been observed, but their full-length nature is not clearly defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: card 9; CARD9; CARD9-N; caspase recruitment domain protein 9; rCARD9; RP11-413M3.9; unnamed protein product
Gene Aliases: CANDF2; Gm782; hCARD9; IMD103
UniProt ID: (Human) Q9H257, (Rat) Q9EPY0, (Mouse) A2AIV8
Entrez Gene ID: (Human) 64170, (Rat) 64171, (Mouse) 332579
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support