Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MYSQRFGTVQ REVKGPTPKV VIVRSKPPKG QGAEHHLERI RRSHQKHNAI LASIKSSERD RLKAEWDQHN DCKILDSLVR ARIKDAVQGF IINIEERRNK LRELLALEEN EYFTEMQLKK ETIEEKKDRM REKTKLLKEK NEKERQDFVA EKLDQQFRER CEELRVELLS IHQKKVCEER KAQIAFNEEL SRQKLVEEQM FSKLWEEDRL AKEKREAQEA RRQKELMENT RLGLNAQITS IKAQRQATQL LKEEEARLVE SNNAQIKHEN EQDMLKKQKA KQETRTILQK ALQERIEHIQ; Positive Samples: Mouse testis, Rat testis; Cellular Location: Cell projection, cilium
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning. [provided by RefSeq, Aug 2015]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 11; coiled-coil domain-containing protein 11; unnamed protein product
Gene Aliases: 4933415I03Rik; CCDC11; HTX6; RGD1306734
UniProt ID: (Human) Q96M91, (Mouse) Q9D439
Entrez Gene ID: (Human) 220136, (Mouse) 74453, (Rat) 364899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support