Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
CCDC83 (Coiled-Coil Domain Containing 83) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain-containing protein 83; unnamed protein product
Gene Aliases: 4930549K11Rik; 4930554C01Rik; 4932423M01Rik; CT148; HSD9; KP-CoT-23
UniProt ID: (Human) Q8IWF9, (Mouse) Q9D4V3
Entrez Gene ID: (Human) 220047, (Mouse) 75338, (Rat) 680270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support