Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Description: This R2.477 monoclonal antibody reacts with human CD112 (also known as Nectin-2 or Poliovirus Receptor Related 2), a 60-65 kDa immunoglobulin-like transmembrane cell adhesion molecule. This cell surface molecule is expressed on hematopoietic cells, endothelial, epithelial, as well as tumor cell lines such as Jurkat, U937, and MCF7. CD112 is a ligand for DNAM-1 (CD226), a leukocyte adhesion molecule expressed on natural killer (NK) cells. Interaction between CD112 and DNAM-1 promotes NK cytotoxic activity, leading to tumor cell lysis. In addition to its role in tumor suppression, CD112 plays a cooperative role in cell-cell adhesion, particularly in the formation of adherens junctions between neighboring epithelial cells and fibroblasts. CD112 can form homophilic and heterophilic interactions to regulate contact inhibition of cell movement and proliferation. Finally, CD112 has been reported to be a receptor for herpesvirus entry into cells.
This monoclonal antibody has been reported to block cell adhesion and herpesvirus entry.
Applications Reported: This R2.477 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This R2.477 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser
This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; Nectin cell adhesion molecule 2; Nectin-2; poliovirus receptor-like 2; poliovirus receptor-related 2 (herpesvirus entry mediator B); Poliovirus receptor-related protein 2
Gene Aliases: CD112; HVEB; NECTIN2; PRR2; PVRL2; PVRR2
UniProt ID: (Human) Q92692
Entrez Gene ID: (Human) 5819
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support