Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Description: This OV1F12 (1F12HE) monoclonal antibody reacts with human CD156c, which is also known as a disintegrin and metalloproteinase domain-containing protein (ADAM) 10. This type I transmembrane glycoprotein is expressed ubiquitously on hematopoietic and non-hematopoietic cell types. Studies demonstrate that CD156c expression is regulated by PAX2 in cancer cells. CD156c plays a critical role in hematopoiesis, including lymphocyte development, and autoimmunity by regulating the shedding and secretion of various receptors and chemokines, including Notch, CD23, CXCL16, and the IL-6R. Finally, deregulated CD156c expression has been associated with T-cell acute lymphoblastic lymphoma (T-ALL) and Alzheimer's disease.
Applications Reported: This OV1F12 antibody has been reported for use in intracellular staining followed by flow cytometric analysis.
Applications Tested: This OV1F12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
Proinflammatory cytokine tumor necrosis factor-a (TNF-a) contributes to a variety of inflammatory responses and programmed cell death. Notch receptor and its ligand participate in cell fate decisions during vertebrate development and are associated with several human disorders, including a T-cell lymphoma. TNF-a, notch and its ligand delta are all membrane-bond molecules, which are cleaved by proteases to release mature proteins or functional receptor. ADAM10, a metalloprotease-disintegrin in the family of mammalian ADAM (for a disintegrin and metalloprotease), was recently identified to cleave TNF-a, notch and its ligand delta (1-3). The genes encoding human, mouse, and bovine ADAM10 were recently cloned and designated ADAM 10, kuzbanian (KUZ), and MADM, respectively. ADAM10 mRNA is expressed in a variety of human and bovine tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support