Search Thermo Fisher Scientific
Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
The siglecs are a family of membrane bound lectins (of the immunoglobulin superfamily) that bind sialic acid and mediate cell-cell interactions. Family members include sialoadhesin, CD22 and CD33. Sialoadhesin is a macrophage-specific cell adhesion molecule that is expressed on activated macrophages in chronic inflammation and in tumours.