This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD2 (clone T6.3, isotype IgG2a).
Dual color combination consisting of FITC conjugated and R-PE conjugated monoclonal antibodies mixed in optimal ratio.