Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EPCR Antibodies

          Invitrogen

          CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), Biotin, eBioscience™

          20 Published Figures
          20 References
          View all (74) EPCR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), Biotin, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (20)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 21

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD201 (EPCR) Antibody (13-2012-82) in Flow

          Mouse bone marrow cells were stained with staining buffer (autofluorescence) (left) or Anti-Mouse CD201 (EPCR) Biotin (right), followed by Streptavidin Alexa Fluor 594, and Anti-Mouse CD34 FITC (Product # 11-0341-82), Anti-Mouse Ly-6A/E (Sca-1) PE (Product # 12-5981-82), Anti-Mouse CD117 (c-Kit) PE-Cy7 (Product # 25-1172-82), and anti-mouse lineage APC. Events displayed are Sca-1+, c-kit+ and Lineage-. Images provided courtesy of Hiromitsu Nakauchi. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunocytochemistry (ICC/IF)
          CD201 (EPCR) Antibody in Immunohistochemistry (IHC)
          CD201 (EPCR) Antibody in Immunohistochemistry (IHC)
          CD201 (EPCR) Antibody in Immunohistochemistry (IHC)
          CD201 (EPCR) Antibody in Immunohistochemistry (IHC)
          CD201 (EPCR) Antibody in Immunohistochemistry (IHC)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)
          CD201 (EPCR) Antibody in Flow Cytometry (Flow)

          Product Details

          13-2012-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          Assay-Dependent
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          -
          View 9 publications 9 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 8 publications 8 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Human, Mouse, Non-human primate

          Host/Isotype

          Rat / IgG2b, kappa

          Recommended Isotype Control

          Rat IgG2b kappa Isotype Control (eB149/10H5), Biotin, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          eBio1560 (1560)

          Conjugate

          Biotin Biotin Biotin
          • APC 
          • APC-eFluor 780
          • Functional Grade
          • PE 
          • PerCP-eFluor 710
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_657694

          Product Specific Information

          Description: The eBio1560 monoclonal antibody reacts with mouse Endothelial Protein C Receptor (EPCR, CD201), a 25 kDa Type 1 transmembrane protein expressed on endothelial cells. EPCR exhibits sequence and structural homology with the MHC class I/CD1 family of proteins. EPCR is a ligand for Protein C and plays an important role in augmenting Protein C activation by the thrombin-thrombomodulin complex and in regulating blood coagulation and inflammation. Deletion of EPCR results in embryonic lethality, at least partically due to placental thrombosis.

          Recently, it was demonstrated that EPCR expression identified cells in the bone marrow that are capable of hematopoietic reconstitution activity comparable to hematopoietic stem cells isolated with conventional methods. The eBio1560 monoclonal antibody can be used for the detection of these hematopoietic stem cells, however the eBiomRCR-16 monoclonal antibody should only be used for the detection of CD201 on endothelial cells.

          Applications Reported: This eBio1560 (1560) antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining.

          Applications Tested: This eBio1560 (1560) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest, particµLarly when analyzing expression on endothelial cells where CD201 expression is very high.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Activated protein C receptor; APC receptor; bA42O4.2; CD antigen CD201; CD201; Centrocyclin; Centrosomal protein CCD41; Endothelial cell protein C receptor; Endothelial protein C receptor

          View more View less

          Gene Aliases: AI325044; Ccca; Ccd41; Epcr; Procr

          View more View less

          UniProt ID: (Mouse) Q64695

          View more View less

          Entrez Gene ID: (Mouse) 19124

          View more View less

          Function(s)
          protein C-terminus binding protein-binding activity modulator
          Process(es)
          immune response blood coagulation hemostasis antigen processing and presentation negative regulation of coagulation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline